Cole-Parmer® Disposable, Weighted Polyethylene, PVC, FEP Bailers, and Groundwater Filter
Large opening for easy tie-off, greater water flow, and improved sink rate
  • Individually sealed bailers are purified and rinsed with deionized water before packaging
  • All bailers are weighted for increased sink rate
These disposable bailers feature tapered ends and low-snag cord cleat to reduce hang-ups and snags. The angled nozzle ensures positive check-valve control. Choose from virgin polyethylene construction for strength and durability, PVC for strength and economy of polyethylene with transparent features, or FEP construction for high-purity applications. Double check-valve bailers allow you to sample from a specific depth (point-source sampling).
6,400.00 - ₹125,000.00INR / Case of 24
18 variations of this product are available.
See All Product Options Below
18 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Number of Valves
Availability Pricing
COMPARE
12 Polyethylene 1
₹42,100.00INR / Case of 24
COMPARE
12 Polyethylene 2
₹42,100.00INR / Case of 24
COMPARE
36 Polyethylene 1
₹36,200.00INR / Case of 24
COMPARE
36 Polyethylene 2
₹43,700.00INR / Case of 24
COMPARE
48 Polyethylene 1
₹52,700.00INR / Case of 24
COMPARE
48 Polyethylene 2
₹55,000.00INR / Case of 24
COMPARE
36 FEP 1
₹125,000.00INR / Case of 12
COMPARE
36 FEP 2
₹117,000.00INR / Case of 12
COMPARE
36 Polyethylene 1
₹32,900.00INR / Case of 24
COMPARE
36 Polyethylene 2
₹41,500.00INR / Case of 24
COMPARE
36 FEP 1
₹33,900.00INR / Case of 12
COMPARE
36 FEP 2
₹48,900.00INR / Case of 12
COMPARE
36 Polyethylene 1
₹37,700.00INR / Case of 24
COMPARE
36 Polyethylene 1
₹45,200.00INR / Case of 24
COMPARE
12 CPVC 1
₹39,200.00INR / Case of 24
COMPARE
36 CPVC 1
₹40,700.00INR / Case of 24
COMPARE
48 CPVC 1
₹52,700.00INR / Case of 24
COMPARE
Polyethersulfone
₹6,400.00INR / Case of 1
Showing 1- 18 of 18