Cole-Parmer Quickfilter High Capacity Ground Water Filter Cartridge, 0.45 uM, 1 per Case
Representative image only.
Cole-Parmer Quickfilter High Capacity Ground Water Filter Cartridge, 0.45 uM, 1 per Case
Cole-Parmer | Item: 9915750
Large opening for easy tie-off, greater water flow, and improved sink rate
₹6,400.00INR / Case of 1
₹6,400.00INR / Case of 1
Product Description
Key Features
  • Individually sealed bailers are purified and rinsed with deionized water before packaging
  • All bailers are weighted for increased sink rate
More About this Item
These disposable bailers feature tapered ends and low-snag cord cleat to reduce hang-ups and snags. The angled nozzle ensures positive check-valve control. Choose from virgin polyethylene construction for strength and durability, PVC for strength and economy of polyethylene with transparent features, or FEP construction for high-purity applications. Double check-valve bailers allow you to sample from a specific depth (point-source sampling).
Disposable filter with 0.45 micron polyethersulfone membrane. Filter has bailer interface tube to connect directly to bailer.
Specifications
  • MaterialPolyethersulfone
  • DescriptionQuickfilter High Capacity Ground Water Filter Cartridge, 0.45 uM, 1 per Case