Cole-Parmer® Cardboard Cryogenic Storage Boxes and Dividers
Create customized storage box configurations
  • Snug-fitting lid prevents accidental separation if dropped
  • Use in mechanical freezers or liquid nitrogen tanks to –196°C
  • Constructed of sturdy cardboard with moisture-repellent coating
  • Designed with small holes on the bottom to allow for drainage
These economical cryo/freezer storage boxes offer excellent low-temperature storage. Moisture-repellent coating protects boxes from effects of liquid nitrogen or condensation.
Choose from a wide range of boxes and dividers to accommodate tubes from 0.5 to 50 mL. Boxes can be ordered with dividers for a complete box configuration or without dividers to customize your storage needs. The wide range of dividers can hold from 16 to 196 tubes with diameters from 8 to 33.5 mm.
Order additional and replacement cell dividers separately.
300.00 - ₹3,000.00INR / Each
29 variations of this product are available.
See All Product Options Below
29 PRODUCT OPTIONS
Compareitem specifications
Tube Size Description
Availability Pricing
COMPARE
0.2 mL PCR Cardboard Freezer Box, 3" x 3" x 1"; with 64-Place Divider for PCR Tubes
₹440.00INR / Each
COMPARE
0.2 mL PCR Cardboard Freezer Box, 5-1/4" x 5-1/4" x 1"; with 196-Place Divider for PCR Tubes
₹500.00INR / Each
COMPARE
0.5, 1.5, and 2 mL Cardboard Freezer Box, 3" x 3" x 2", with 25-Place Divider
₹400.00INR / Each
COMPARE
0.5, 1.5, and 2 mL Cardboard Freezer Box, 5-1/4" x 5-1/4" x 2", with 81-Place Divider
₹550.00INR / Each
COMPARE
0.5, 1.5, and 2 mL Cardboard Freezer Box, 5-1/4" x 5-1/4" x 2", with 100-Place Divider
₹300.00INR / Each
COMPARE
Not divided Cardboard Freezer Box, 3" x 3" x 2"; without Divider
₹440.00INR / Each
COMPARE
Not divided Cardboard Freezer Box, 5-3/4" x 5-3/4" x 4-7/8"H; without Divider
₹550.00INR / Each
COMPARE
Not divided Cardboard Freezer Box, 5-1/4" x 5-1/4" x 2"; without Divider
₹440.00INR / Each
COMPARE
Not divided Cardboard Freezer Box, 5-1/4" x 5-1/4" x 3"; without Divider
₹440.00INR / Each
COMPARE
50 mL Cardboard Freezer Box Divider, 5-3/8" x 5-3/8" x 2-3/4", 16-Place, for 50 mL Tubes; 12/pk
₹2,300.00INR / Pkg of 12
COMPARE
15 mL Cardboard Freezer Box Divider, 5-3/8" x 5-3/8" x 2-3/4", 36-Place, for 15 mL Tubes; 12/pk
₹2,300.00INR / Pkg of 12
COMPARE
0.5, 1.5, and 2 mL Cardboard Freezer Box Divider, 2-5/8" x 2-5/8" x 1-1/4"; 25-Place
₹2,300.00INR / Pkg of 12
COMPARE
15 and 50 mL Cardboard Freezer Box Divider, 4-7/8" x 4-7/8" x 1-5/16"; 25-Place
₹2,300.00INR / Pkg of 12
COMPARE
1.5 mL and 2.0 mL Cardboard Freezer Box Divider, 4-7/8" x 4-7/8" x 1-5/16"; 49-Place
₹2,100.00INR / Pkg of 12
COMPARE
1.5 mL and 2.0 mL Cardboard Freezer Box Divider, 4-7/8" x 4-7/8" x 1-5/16"; 64-Place
₹2,200.00INR / Pkg of 12
COMPARE
0.5, 1.5, and 2 mL Cardboard Freezer Box Divider, 4-7/8" x 4-7/8" x 1-5/16"; 81-Place
₹2,300.00INR / Pkg of 12
COMPARE
0.5 mL and 1.5 mL Cardboard Freezer Box Divider, 4-7/8" x 4-7/8" x 1-5/16"; 100-Place
₹2,300.00INR / Pkg of 12
COMPARE
0.5 mL and 1.5 mL Cardboard Freezer Box Divider, 4-7/8" x 4-7/8" x 1-5/16"; 144-Place
₹2,300.00INR / Pkg of 12
COMPARE
Not divided Cardboard Freezer Box, Plain White, 5-3/4" x 5-3/4" x 4-7/8"H; without Divider
₹590.00INR / Each
COMPARE
Not divided Cardboard Freezer Box, Plain White, 5-1/4" x 5-1/4" x 2"; without Divider
₹440.00INR / Each
Showing 1 - 20 of 29