CELLTREAT Sterile Tissue Culture Flasks
Angled neck design offers easy access
  • Easily view cells through clear polystyrene
  • Sterilized by gamma irradiation
56,100.00 - ₹75,200.00INR / Case of 200
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap Type Culture Area (cm²)
Availability Pricing
CELLTREAT Scientific Products 229320 Sterile Treated Culture Flasks with Plug Seal Cap, 12.5 cm²; 200/cs 0193101 Mfr # 229320
COMPARE
25 Plug seal 12.5
₹66,500.00INR / Case of 200
CELLTREAT Scientific Products 229321 Sterile Treated Culture Flasks with Vented Cap, 12.5 cm²; 200/cs 0193102 Mfr # 229321
COMPARE
25 Vent 12.5
₹71,200.00INR / Case of 200
CELLTREAT Scientific Products 229330 Sterile Treated Culture Flasks with Plug Seal Cap, 25 cm²; 200/cs 0193103 Mfr # 229330
COMPARE
50 Plug seal 25
₹70,500.00INR / Case of 200
CELLTREAT Scientific Products 229331 Sterile Treated Culture Flasks with Vented Cap, 25 cm²; 200/cs 0193104 Mfr # 229331
COMPARE
50 Vent 25
₹75,200.00INR / Case of 200
CELLTREAT Scientific Products 229340 Sterile Treated Culture Flasks with Plug Seal Cap, 75 cm², 100/cs 0193105 Mfr # 229340
COMPARE
250 Plug seal 75
₹68,300.00INR / Case of 100
CELLTREAT Scientific Products 229341 Sterile Treated Culture Flasks with Vented Cap, 75 cm²; 100/cs 0193106 Mfr # 229341
COMPARE
250 Vent 75
₹72,000.00INR / Case of 100
CELLTREAT Scientific Products 229350 Sterile Treated Culture Flasks with Plug Seal Cap, 182 cm²; 40/cs 0193107 Mfr # 229350
COMPARE
600 Plug seal 182
₹58,200.00INR / Case of 40
CELLTREAT Scientific Products 229351 Sterile Treated Culture Flasks with Vented Cap, 182 cm²; 40/cs 0193108 Mfr # 229351
COMPARE
600 Vent 182
₹61,000.00INR / Case of 40
CELLTREAT Scientific Products 229361 Sterile Treated Culture Flasks with Vented Caps, 300 cm²; 18/cs 0193109 Mfr # 229361
COMPARE
1250 Plug seal 300
₹65,700.00INR / Case of 18
CELLTREAT Scientific Products 229500 Sterile Suspension Culture Flasks with Vented Cap, 25 mL; 200/cs 0193111 Mfr # 229500
COMPARE
25 Vent
₹62,200.00INR / Case of 200
CELLTREAT Scientific Products 229510 Sterile Suspension Culture Flasks with Vented Cap, 50 mL; 200/cs 0193112 Mfr # 229510
COMPARE
50 Vent
₹66,000.00INR / Case of 200
CELLTREAT Scientific Products 229520 Sterile Suspension Culture Flasks with Vented Cap, 250 mL; 100/cs 0193114 Mfr # 229520
COMPARE
250 Vent
₹61,500.00INR / Case of 100
CELLTREAT Scientific Products 229530 Sterile Suspension Culture Flasks with Vented Cap, 600 mL; 40/cs 0193116 Mfr # 229530
COMPARE
600 Vent
₹56,100.00INR / Case of 40
Showing 1- 13 of 13