CELLTREAT Scientific Products 229330 Sterile Treated Culture Flasks with Plug Seal Cap, 25 cm²; 200/cs
Representative image only.
CELLTREAT Scientific Products 229330 Sterile Treated Culture Flasks with Plug Seal Cap, 25 cm²; 200/cs
CELLTREAT Scientific Products | Mfr: 229330| Item: 0193103
Angled neck design offers easy access
₹70,500.00INR / Case of 200
₹70,500.00INR / Case of 200
Buy It Together
Current Item
CELLTREAT Scientific Products 229330 Sterile Treated Culture Flasks with Plug Seal Cap, 25 cm²; 200/cs
CELLTREAT Scientific Products 229330 Sterile Treated Culture Flasks with Plug Seal Cap, 25 cm²; 200/cs
0193103
₹70,500.00INR / Case of 200
+ =
Total Price 74100.00 ADD 2 ITEMS TO CART
Product Description
Includes
Sterile plug seal caps.
Key Features
  • Easily view cells through clear polystyrene
  • Sterilized by gamma irradiation
  • Hydrophilic surface is ideal for consistent cell attachment
Sterile
Specifications
  • MaterialPolystyrene
  • Capacity (mL)50
  • Cap TypePlug seal
  • Culture Area (cm²)25
  • DescriptionSterile Treated Culture Flasks with Plug Seal Cap, 25 cm²; 200/cs