CELLTREAT Scientific Products 229390 Vented Cap for 25 cm² and 50 mL Sterile Culture Flasks; 5/cs
Representative image only.
CELLTREAT Scientific Products 229390 Vented Cap for 25 cm² and 50 mL Sterile Culture Flasks; 5/cs
CELLTREAT Scientific Products | Mfr: 229390| Item: 0193123
Fits CellTreat Treated Culture Flasks and Untreated Solution Flasks
₹4,100.00INR / Case of 5
₹4,100.00INR / Case of 5
Product Description
Key Features
  • Sterilized by gamma irradiation
  • Designed to fit 50-mL cell culture flasks
  • Vented caps feature 0.22 µm hydrophobic membrane
More About this Item
Choose from plug seal and vent cap types. Culture flasks are sold separately.
Sterile
Specifications
  • Cap TypeVent
  • DescriptionVented Cap for 25 cm² and 50 mL Sterile Culture Flasks; 5/cs