CELLTREAT Scientific Products 229396 Plug Seal Cap for 75 cm² and 250 mL Sterile Culture Flasks; 5/cs
Representative image only.
CELLTREAT Scientific Products 229396 Plug Seal Cap for 75 cm² and 250 mL Sterile Culture Flasks; 5/cs
CELLTREAT Scientific Products | Mfr: 229396| Item: 0193129
Fits CellTreat Treated Culture Flasks and Untreated Solution Flasks
₹3,600.00INR / Case of 5
₹3,600.00INR / Case of 5
Product Description
Key Features
  • Sterilized by gamma irradiation
  • Designed to fit 250-mL cell culture flasks
More About this Item
Choose from plug seal and vent cap types. Culture flasks are sold separately.
Sterile
Specifications
  • Cap TypePlug seal
  • DescriptionPlug Seal Cap for 75 cm² and 250 mL Sterile Culture Flasks; 5/cs