CELLTREAT Scientific Products 229350 Sterile Treated Culture Flasks with Plug Seal Cap, 182 cm²; 40/cs
Representative image only.
CELLTREAT Scientific Products 229350 Sterile Treated Culture Flasks with Plug Seal Cap, 182 cm²; 40/cs
CELLTREAT Scientific Products | Mfr: 229350| Item: 0193107
Angled neck design offers easy access
₹58,200.00INR / Case of 40
₹58,200.00INR / Case of 40
Buy It Together
Current Item
CELLTREAT Scientific Products 229350 Sterile Treated Culture Flasks with Plug Seal Cap, 182 cm²; 40/cs
CELLTREAT Scientific Products 229350 Sterile Treated Culture Flasks with Plug Seal Cap, 182 cm²; 40/cs
0193107
₹58,200.00INR / Case of 40
+ =
Total Price 62300.00 ADD 2 ITEMS TO CART
Product Description
Includes
Sterile plug seal caps.
Key Features
  • Easily view cells through clear polystyrene
  • Sterilized by gamma irradiation
  • Hydrophilic surface is ideal for consistent cell attachment
  • Thicker, more durable walls for improved stacking capability
  • Printed lot numbers
Sterile
Specifications
  • MaterialPolystyrene
  • Capacity (mL)600
  • Cap TypePlug seal
  • Culture Area (cm²)182
  • DescriptionSterile Treated Culture Flasks with Plug Seal Cap, 182 cm²; 40/cs