CELLTREAT Scientific Products 229340 Sterile Treated Culture Flasks with Plug Seal Cap, 75 cm², 100/cs
Representative image only.
CELLTREAT Scientific Products 229340 Sterile Treated Culture Flasks with Plug Seal Cap, 75 cm², 100/cs
CELLTREAT Scientific Products | Mfr: 229340| Item: 0193105
Angled neck design offers easy access
₹68,300.00INR / Case of 100
₹68,300.00INR / Case of 100
Buy It Together
Current Item
CELLTREAT Scientific Products 229340 Sterile Treated Culture Flasks with Plug Seal Cap, 75 cm², 100/cs
CELLTREAT Scientific Products 229340 Sterile Treated Culture Flasks with Plug Seal Cap, 75 cm², 100/cs
0193105
₹68,300.00INR / Case of 100
+ =
Total Price 71900.00 ADD 2 ITEMS TO CART
Product Description
Includes
Sterile plug seal caps.
Key Features
  • Easily view cells through clear polystyrene
  • Sterilized by gamma irradiation
  • Hydrophilic surface is ideal for consistent cell attachment
  • Thicker, more durable walls for improved stacking capability
  • Printed lot numbers
Sterile
Specifications
  • MaterialPolystyrene
  • Capacity (mL)250
  • Cap TypePlug seal
  • Culture Area (cm²)75
  • DescriptionSterile Treated Culture Flasks with Plug Seal Cap, 75 cm², 100/cs