Accessories and Parts for Advantec Polysulfone Filter Holders
Enhance the life of your Advantec Polysulfone filter holders
These parts and accesories are suitable for use with your Advantec polysulfone filter holders.
The funnel cover can be vented aseptically using disposable syringe filter.
The recessed filter support provides sufficient clearance so that the membrane will not twist or tear when the funnel is secured.
Aseptic Polysulfone Filter Holder sold separately as part numbers 06644-70, -72, -74, and -76.
22,600.00 - ₹64,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Advantec 311401 300ML FUNNEL KG47 0664533 Mfr # 311401
COMPARE
₹64,600.00INR / Each
Advantec 501002-2PK Funnel for Polysulfone Filter Holders; 300 mL capacity 0664588 Mfr # 44501002
COMPARE
Funnel for Polysulfone Filter Holders; 300 mL capacity
₹64,800.00INR / Pkg of 1
Advantec 501004-A Polypropylene Support Screen for Polysulfone Filter Holders 0664590 Mfr # 44501004
COMPARE
Polypropylene Support Screen for Polysulfone Filter Holders
₹22,600.00INR / Pkg of 4