Advantec 501004-A Polypropylene Support Screen for Polysulfone Filter Holders
Representative image only.
Advantec 501004-A Polypropylene Support Screen for Polysulfone Filter Holders
Advantec | Mfr: 44501004| Item: 0664590
Enhance the life of your Advantec Polysulfone filter holders
₹22,600.00INR / Pkg of 4
₹22,600.00INR / Pkg of 4
Buy It Together
Current Item
Advantec 501004-A Polypropylene Support Screen for Polysulfone Filter Holders
Advantec 501004-A Polypropylene Support Screen for Polysulfone Filter Holders
0664590
₹22,600.00INR / Pkg of 4
+ =
Total Price 34000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
These parts and accesories are suitable for use with your Advantec polysulfone filter holders.
The funnel cover can be vented aseptically using disposable syringe filter.
The recessed filter support provides sufficient clearance so that the membrane will not twist or tear when the funnel is secured.
Aseptic Polysulfone Filter Holder sold separately as part numbers 06644-70, -72, -74, and -76.
1 Year Warranty
Specifications
  • DescriptionPolypropylene Support Screen for Polysulfone Filter Holders