Advantec® Polypropylene Holders
Aseptic sampling of liquids and gases
  • Maximum operating temperature (liquids) is 176°F (80°C)
  • Holders feature polypropylene dual support screens and silicone O-rings
Use these polypropylene (PP) holders for ultracleaning and sterilizing liquids under pressure, or for aseptic sampling of liquids and gases. Dual support screens allow for flow in both directions. Assemble holders quickly and efficiently without membrane tears with the specially designed inlet cap and exterior lock.
11,400.00 - ₹17,500.00INR / Pkg of 6
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Filter Diameter (mm)
Availability Pricing
Advantec 43303020 Polypropylene Filter Holder for 47 mm Membranes 0662322 Mfr # 43303020
COMPARE
Polypropylene 47
₹11,400.00INR / Each
Advantec 43303010 Polypropylene Filter Holder for 25 mm Membranes 0662332 Mfr # 43303010
COMPARE
Polypropylene 25
₹17,500.00INR / Pkg of 6