Advantec 311401 300ML FUNNEL KG47
Representative image only.
Advantec 311401 300ML FUNNEL KG47
Advantec | Mfr: 311401| Item: 0664533
Enhance the life of your Advantec Polysulfone filter holders
₹64,600.00INR / Each
₹64,600.00INR / Each
Buy It Together
Current Item
Advantec 311401 300ML FUNNEL KG47
Advantec 311401 300ML FUNNEL KG47
0664533
₹64,600.00INR / Each
+ =
Total Price 76000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
These parts and accesories are suitable for use with your Advantec polysulfone filter holders.
The funnel cover can be vented aseptically using disposable syringe filter.
The recessed filter support provides sufficient clearance so that the membrane will not twist or tear when the funnel is secured.
Aseptic Polysulfone Filter Holder sold separately as part numbers 06644-70, -72, -74, and -76.