Advantec 501002-2PK Funnel for Polysulfone Filter Holders; 300 mL capacity
Representative image only.
Advantec 501002-2PK Funnel for Polysulfone Filter Holders; 300 mL capacity
Advantec | Mfr: 44501002| Item: 0664588
Enhance the life of your Advantec Polysulfone filter holders
₹64,800.00INR / Pkg of 1
₹64,800.00INR / Pkg of 1
Buy It Together
Current Item
Advantec 501002-2PK Funnel for Polysulfone Filter Holders; 300 mL capacity
Advantec 501002-2PK Funnel for Polysulfone Filter Holders; 300 mL capacity
0664588
₹64,800.00INR / Pkg of 1
+ =
Total Price 76200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
More About this Item
These parts and accesories are suitable for use with your Advantec polysulfone filter holders.
The funnel cover can be vented aseptically using disposable syringe filter.
The recessed filter support provides sufficient clearance so that the membrane will not twist or tear when the funnel is secured.
Aseptic Polysulfone Filter Holder sold separately as part numbers 06644-70, -72, -74, and -76.
1 Year Warranty
Specifications
  • Body MaterialPolysulfone
  • DescriptionFunnel for Polysulfone Filter Holders; 300 mL capacity