Abdos Large-Capacity Polypropylene Beakers
No need to mix multiple smaller volumes
  • Constructed of durable polypropylene
  • Autoclavable
Large-capacity polypropylene (PP) beakers are ideal for general laboratory use. Beakers are tapered for easy stacking, saves critical lab space! Screened-on black graduations are easy to read and provide approximate graduation reading. Available with or without handles. Withstand temperatures from 32 to 250°F (0 to 121°C).
14,500.00 - ₹20,800.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Material
Availability Pricing
Abdos P50607 Large Capacity PP Beakers, 5000 mL, 2/Pk 6100051 Mfr # P50607
COMPARE
5000 500 Polypropylene(PP)
₹14,500.00INR / Pkg of 2
Abdos P50608 Large Capacity PP Beaker; 10,000 mL, 1/Pk 6100053 Mfr # P50608
COMPARE
10000 1000 Polypropylene(PP)
₹18,300.00INR / Each
Abdos P50807 Large Capacity Beaker with Handle, PP, 5L, 2 pack 6100055 Mfr # P50807
COMPARE
5000 500 Polypropylene(PP)
₹20,800.00INR / Pkg of 2
Abdos P50808 Large Capacity Beaker with Handle, PP, 10L, 1 pack 6100057 Mfr # P50808
COMPARE
10000 1000 Polypropylene(PP)
₹19,300.00INR / Each