A&D Weighing Borealis BA Series Micro and Semi-Micro Analytical Balances
Enhanced functions for effortless weighing
  • External ionizer eliminates static electricity before weighing
  • IR sensors provide touchless opening and closing of door
  • Perform automatic calibration adjustment with internal weight
  • A 5" remote touch-screen display for easy operation
  • Effortless leveling with on-screen leveling assist
  • Easy-to-clean removable draft shield
  • Built-in clock for output weighing value with time/date stamp
  • Password protection prevents accidental setting changes
  • Comes with underhook for weighing magnetic materials
  • Easily connect to external devices using RS-232, USB-B, USB-A, Ethernet TCP/IP, Bluetooth®, and two input ports
  • GLP/GMP/GCP/ISO compliant output
The Borealis BA Series micro and semi-micro analytical balances come fully equipped to provide accurate and precise measurements. Balances offer capacities from 6.2 to 120 g with four different weighing applications—normal weighing, counting mode, percent weighing, minimum weight alert function. Antistatic glass panels along with an external ionizer ensure static electricity is eliminated before weighing to reduce errors. A self-sensitivity adjustment feature uses the internal weight to automatically calibrate according to the set execution conditions (temperature change, preset time, or preset interval) when there is nothing on the pan to ensure consistent measurement results.
Control balance functions and view measurements on the large remote display unit. Remote display ensures weighing unit is not disturbed while the measurement is being taken. Display unit features a 5" LCD, resistive color screen; two IR sensors; and four quick access keys for easy navigation. Touchscreen enables intuitive operation while making it easy to enter numbers and change settings, even while wearing thick gloves. Easily level balance using the on-screen leveling assist feature with visual instructions indicating which foot to rotate. Program IR sensors to activate functions such as auto door open/close, print, and zero. Draft shield door can be set to open fully or partially depending on your preference.
The glass panels on the draft shield use a unique clip system for fast and easy removal. Sliding doors hang from the top of the housing so there are no grooves where contaminants can accumulate. To clean, simply unclip the glass panels or remove the draft shield by pulling out the fasteners.
1,631,000.00 - ₹4,386,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
A&D Weighing BA-6TE Borealis BA Series Micro Analytical Balance, 6.2g x 0.001mg 5500051 Mfr # BA-6TE
COMPARE
6.2 0.001 Micro Analytical Balance, 6.2g x 0.001mg
₹4,386,000.00INR / Each
A&D Weighing BA-6DTE Borealis BA Series Micro Analytical Balance, 2.1g x 0.001mg / 6.2g x 0.01mg 5500052 Mfr # BA-6DTE
COMPARE
6.2 0.01 Micro Analytical Balance, 2.1g x 0.001mg / 6.2g x 0.01mg
₹4,079,000.00INR / Each
A&D Weighing BA-225TE Borealis BA Series Semi-Micro Analytical Balance, 220g x 0.01mg 5500053 Mfr # BA-225TE
COMPARE
220 0.01 Semi-Micro Analytical Balance, 220g x 0.01mg
₹2,031,000.00INR / Each
A&D Weighing BA-225DTE Borealis BA Series Semi-Micro Analytical Balance, 51g x 0.01mg / 220g x 0.1mg 5500054 Mfr # BA-225DTE
COMPARE
220 0.1 Semi-Micro Analytical Balance, 51g x 0.01mg / 220g x 0.1mg
₹1,835,000.00INR / Each
A&D Weighing BA-125DTE Borealis BA Series Semi-Micro Analytical Balance, 51g x 0.01mg / 120g x 0.1mg 5500055 Mfr # BA-125DTE
COMPARE
120 0.1 Semi-Micro Analytical Balance, 51g x 0.01mg / 120g x 0.1mg
₹1,631,000.00INR / Each