A-310 High-Efficiency Axial Flow Impellers
Offers 50% more mixing action
  • Maximizes flow with ability to add shear options
High-efficiency axial flow impellers have laser designed paddles to develop 50% more mixing action for high-flow, low-shear mixing. Recommended for low-viscosity flow-controlled applications.
40,200.00 - ₹209,000.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
Compareitem specifications
Bore Diameter (mm) Diameter (mm)
Availability Pricing
COMPARE
10 86
₹47,500.00INR / Each
COMPARE
8 96
₹45,200.00INR / Each
COMPARE
8 114
₹46,900.00INR / Each
COMPARE
8 64
₹40,200.00INR / Each
808956 High-Efficiency Axial Flow Impeller, 3/8" Bore; 2.5" Dia 0456050 Mfr # 808956
COMPARE
10 64
₹81,100.00INR / Each
871581PSP High-Efficiency Axial Flow Impeller, 5/8" Bore; 3.8" Dia 5035040 Mfr # 871581PSP
COMPARE
16 96
₹110,000.00INR / Each
808887PSP High-Efficiency Axial Flow Impeller, 5/8" Bore; 4.5" Dia 5035042 Mfr # 808887PSP
COMPARE
16 114
₹117,000.00INR / Each
831180PSP High-Efficiency Axial Flow Impeller, 3/4" Bore; 8.8" Dia 5035044 Mfr # 831180PSP
COMPARE
19 224
₹153,000.00INR / Each
809241PSP High-Efficiency Axial Flow Impeller, 3/4" Bore; 10" Dia 5035046 Mfr # 809241PSP
COMPARE
19 254
₹167,000.00INR / Each
808911PSP High-Efficiency Axial Flow Impeller, 3/4" Bore; 11.8" Dia 5035048 Mfr # 808911PSP
COMPARE
19 300
₹188,000.00INR / Each
871235 High-Efficiency Axial Flow Impeller, 3/4" Bore; 7.6" Dia 5035082 Mfr # 871235
COMPARE
19 193
₹143,000.00INR / Each
808959PSP High-Efficiency Axial Flow Impeller, 3/4" Bore; 11.2" Dia 5035084 Mfr # 808959PSP
COMPARE
19 284
₹181,000.00INR / Each
808961 High-Efficiency Axial Flow Impeller, 3/4" Bore; 12.8" Dia 5035086 Mfr # 808961
COMPARE
19 325
₹209,000.00INR / Each
871230PSP High-Efficiency Axial Flow Impeller, 3/4" Bore; 6.3" Dia 5035088 Mfr # 871230PSP
COMPARE
19 160
₹137,000.00INR / Each
Showing 1- 14 of 14