YSI 605203 Replacement Pro Series Polarographic DO Probe with Cap Membrane Kit; 1.25 mil PE
Representative image only.
YSI 605203 Replacement Pro Series Polarographic DO Probe with Cap Membrane Kit; 1.25 mil PE
YSI | Mfr: 605203| Item: 5935234
Optimize the performance of YSI meters
₹71,600.00INR / Each
₹71,600.00INR / Each
Buy It Together
Current Item
YSI 605203 Replacement Pro Series Polarographic DO Probe with Cap Membrane Kit; 1.25 mil PE
YSI 605203 Replacement Pro Series Polarographic DO Probe with Cap Membrane Kit; 1.25 mil PE
5935234
₹71,600.00INR / Each
+
YSI Pro30™ Conductivity Meter
5935205
₹200,000.00INR / Each
+ +
YSI Pro1030 Waterproof Handheld pH/Conductivity Meter
5935105
₹287,000.00INR / Each
+
YSI PRO 10 Pro 10 pH/ORP/temperature portable meter
5935216
₹191,000.00INR / Each
+ + +
YSI Pro 1020 pH and Dissolved Oxygen Meter
5935210
₹287,000.00INR / Each
+
YSI 6050020 Pro20 Dissolved Oxygen Meter, waterproof
5935204
₹187,000.00INR / Each
+
YSI Pro20i Dissolved Oxygen Meter, 1-m Cable, Polarographic
5935340
₹301,000.00INR / Each
=
Total Price 1953900.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Cap membrane kit with 6 yellow screw-on cap membranes (1.25 mil PE), a bottle of electrolyte, and 1 sanding disc.
Key Features
  • Choose from a variety of accessories
  • Easy replace probe for Pro20, Pro20i, Pro1020, Pro2030, and Prop Plus meters—no tools necessary
  • Convenient screw-on cap membranes make membrane changes quick and easy
  • 1 year warranty
More About this Item
Replacement electrodes, cables, power adapters, and other accessories for your YSI water analysis meters.
1 Year Warranty
Specifications
  • Compatible WithYSI Professional Plus
  • DescriptionPro Series Replacement DO Probe; Polarographic