YSI 5906 DO Cap Membrane Kit; 6 Black Screw-on Cap Membranes (1 mil PTFE), Bottle of Electrolyte, and Sanding Disc
Representative image only.
YSI 5906 DO Cap Membrane Kit; 6 Black Screw-on Cap Membranes (1 mil PTFE), Bottle of Electrolyte, and Sanding Disc
YSI | Mfr: 059880| Item: 0551961
Makes membrane chanages quick and easy
₹23,000.00INR / Each
₹23,000.00INR / Each
Buy It Together
Current Item
YSI 5906 DO Cap Membrane Kit; 6 Black Screw-on Cap Membranes (1 mil PTFE), Bottle of Electrolyte, and Sanding Disc
YSI 5906 DO Cap Membrane Kit; 6 Black Screw-on Cap Membranes (1 mil PTFE), Bottle of Electrolyte, and Sanding Disc
0551961
₹23,000.00INR / Each
+
YSI 605101 Pro Series Replacement pH Electrode
5935220
₹55,200.00INR / Each
=
Total Price 78200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Six black screw-on cap membranes (1 mil PTFE), a bottle of electrolyte, and 1 sanding disc.
Key Features
  • Teflon caps offer reliable performance for most dissolved oxygen applications
  • Provides superb fouling resistance
  • 1 year warranty
More About this Item
The DO cap membrane kit is ideal for use with the YSI DO probes and DO meters. It features a fouling resistance with a response time of 18 seconds and flow dependence of 60%.
1 Year Warranty
Specifications
  • Compatible With YSI 5239, 5905, 5010 DO Probes and the 556 and 85 DO meters
  • DescriptionReplacement DO membrane cap kit, 1.00 mil PTFE