YSI 5909 DO Cap Membrane Kit; 6 Blue Screw-on Cap Membranes (2 mil PE), Bottle of Electrolyte, and Sanding Disc
Representative image only.
YSI 5909 DO Cap Membrane Kit; 6 Blue Screw-on Cap Membranes (2 mil PE), Bottle of Electrolyte, and Sanding Disc
YSI | Mfr: 605307| Item: 9985282
Optimize the performance of YSI meters
₹22,300.00INR / Each
₹22,300.00INR / Each
Buy It Together
Current Item
YSI 5909 DO Cap Membrane Kit; 6 Blue Screw-on Cap Membranes (2 mil PE), Bottle of Electrolyte, and Sanding Disc
YSI 5909 DO Cap Membrane Kit; 6 Blue Screw-on Cap Membranes (2 mil PE), Bottle of Electrolyte, and Sanding Disc
9985282
₹22,300.00INR / Each
+
YSI Pro30™ Conductivity Meter
5935205
₹200,000.00INR / Each
+ +
YSI Pro1030 Waterproof Handheld pH/Conductivity Meter
5935105
₹287,000.00INR / Each
+
YSI PRO 10 Pro 10 pH/ORP/temperature portable meter
5935216
₹191,000.00INR / Each
+ + +
YSI Pro 1020 pH and Dissolved Oxygen Meter
5935210
₹287,000.00INR / Each
+
YSI 6050020 Pro20 Dissolved Oxygen Meter, waterproof
5935204
₹187,000.00INR / Each
+
YSI Pro20i Dissolved Oxygen Meter, 1-m Cable, Polarographic
5935340
₹301,000.00INR / Each
=
Total Price 1904600.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Cap membrane kit with 6 blue screw-on cap membranes (2 mil PE), a bottle of electrolyte, and 1 sanding disc.
Key Features
  • Choose from a variety of accessories
  • Convenient screw-on cap membranes make membrane changes quick and easy
  • Blue cap membranes
More About this Item
Replacement electrodes, cables, power adapters, and other accessories for your YSI water analysis meters.
Specifications
  • Compatible WithYSI 556 multiparamter meter
  • DescriptionDO Membrane Kit, 2.00 mil PE for low flow, profiling and flow cell applications