YSI 2003B Replacement Pro Series Polarographic Dissolved Oxygen Sensor with 5909 DO Membrane Kit
Representative image only.
YSI 2003B Replacement Pro Series Polarographic Dissolved Oxygen Sensor with 5909 DO Membrane Kit
YSI | Mfr: 607203| Item: 5935346
Optimize the performance of YSI meters
₹59,600.00INR / Each
₹59,600.00INR / Each
Buy It Together
Current Item
YSI 2003B Replacement Pro Series Polarographic Dissolved Oxygen Sensor with 5909 DO Membrane Kit
YSI 2003B Replacement Pro Series Polarographic Dissolved Oxygen Sensor with 5909 DO Membrane Kit
5935346
₹59,600.00INR / Each
+
YSI Pro30™ Conductivity Meter
5935205
₹200,000.00INR / Each
+ +
YSI Pro1030 Waterproof Handheld pH/Conductivity Meter
5935105
₹287,000.00INR / Each
+
YSI PRO 10 Pro 10 pH/ORP/temperature portable meter
5935216
₹191,000.00INR / Each
+ + +
YSI Pro 1020 pH and Dissolved Oxygen Meter
5935210
₹287,000.00INR / Each
+
YSI 6050020 Pro20 Dissolved Oxygen Meter, waterproof
5935204
₹187,000.00INR / Each
+
YSI Pro20i Dissolved Oxygen Meter, 1-m Cable, Polarographic
5935340
₹301,000.00INR / Each
=
Total Price 1941900.00 ADD 10 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Cap membrane kit with 6 blue screw-on cap 2 mil PE membranes, a bottle of electrolyte, and 1 sanding disc.
Key Features
  • Choose from a variety of accessories
  • Easy replace probe for Pro20, Pro20i, Pro1020, Pro2030, and Prop Plus meters—no tools necessary
  • Convenient screw-on cap membranes make membrane changes quick and easy
  • 3 year warranty
More About this Item
Replacement electrodes, cables, power adapters, and other accessories for your YSI water analysis meters.
3 Year Warranty
Specifications
  • Compatible WithYSI Professional Plus