Traceable Wi-Fi Data Logging Ultra-Low Temperature Thermometer Bundle, 1 Stainless Steel Probe​, with 1-Year Standard TraceableLIVE® Subscription
Representative image only.
Traceable Wi-Fi Data Logging Ultra-Low Temperature Thermometer Bundle, 1 Stainless Steel Probe​, with 1-Year Standard TraceableLIVE® Subscription
Traceable | Mfr: 6656TR| Item: 6500261
One year standard TraceableLIVE® subscription included with purchase
₹90,900.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹90,900.00INR / Each
Buy It Together
Current Item
Traceable Wi-Fi Data Logging Ultra-Low Temperature Thermometer Bundle, 1 Stainless Steel Probe​, with 1-Year Standard TraceableLIVE® Subscription
Traceable Wi-Fi Data Logging Ultra-Low Temperature Thermometer Bundle, 1 Stainless Steel Probe​, with 1-Year Standard TraceableLIVE® Subscription
6500261
₹90,900.00INR / Each
+ + + + + =
Total Price 308200.00 ADD 6 ITEMS TO CART
Product Description
Includes
Traceable® Certificate, 1-year TraceableLIVE® subscription, and thermometer with probe.
Key Features
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • TraceableLIVE-capable products securely monitor your critical environments and notify your smartphone, tablet, or PC if anything goes wrong
  • Notifications for peace of mind - everyone on your team can receive e-mail, text, and push notifications on the environments you monitor
  • Remotely view data logging history, effortlessly generate reports, and control alarm parameters
  • Digital data reports sent with ease whenever you need them
  • Customize your report with location, device, date range, and report format
  • 1 year warranty
More About this Item
Order a thermometer for your unique application and get a one-year standard TraceableLIVE® subscription included.
A standard subscription includes the following features: E-mail and Push Alarms, Unlimited Data Storage, Unlimited Devices, Unlimited Team Members, and Calibration Due notification. Optional SMS text notifications are not included with the subscription code, but can be added on for (+2$/device/month) in the TraceableLIVE® app.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
Guaranteed to have at least one year of calibration remaining on certificate.
Conformite Europeenne1 Year WarrantyNIST Traceable Calibration
Specifications
  • Number of Channels1
  • Cloud ConnectedYes
  • Input TypeOne stainless steel RTD probe
  • Max Temperature (° F)221
  • Min Temperature (° F)-130
  • Min Temperature (° C)-90
  • Max Temperature (° C)105
  • Temperature Resolution (° F)0.01
  • Temperature Accuracy (° F)±0.18
  • Temperature Resolution (° C)0.01
  • Temperature Accuracy (° C)±0.1
  • Sampling Rate12 seconds for data logging; 15 minutes default Wi-Fi transmission
  • Memory Size672 records (7 days worth if set to 15 minute interval)
  • InterfaceWi-Fi
  • System RequirementsMeter and TraceableLIVE subscription
  • Alarm100 (stored max)
  • ConnectionsUSB
  • Battery Type4 AAA (included)
  • Display TypeDual-line LCD
  • DescriptionWi-Fi Datalogging Ultra-Low Temperature Thermometer Bundle, 1 Stainless Steel Probe​, with 1-Year Standard TraceableLIVE® Subscription