Traceable® Memory-Loc™ Data Logging Thermometers with Calibration
Raw data is locked and may not be cleared or changed on base unit
  • Over one million data point capacity!
  • Store up to 10 unique alarm events
  • Smart-Alarm™ with visual/audible alarm continues to alert even if the unit returns to nonalarm conditions
  • Triple-display—shows high, low, and current temperature simultaneously
  • Can upload a USB downloaded file and store on a TraceableLIVE® cloud account
Easily monitor temperatures overnight or for any extended time period with the Memory-Loc datalogging thermometer. Set logging intervals ranging from 1 min to 24 hours to capture over one million temperature data points. Datalogger will stop recording when storage is full and will not overwrite recorded data—perfect for use in 21 CFR 11 environments.
Set high and low alarms in 1° increments; alarm sounds when temperature rises above or falls below set points. The alarm tracking feature stores data for up to 10 different alarm events detailing when the alarm state occurred, as well as when the unit returned within range. Alarm state indicators include audio alerts and flashing LCD segments. Transfer data recorded in a CSV format to any computer via a USB flash drive—no software is required. Unit continues to record data while downloading data.
Status indicators include low-battery, memory full, USB data transfer, and active alarm state. Probe is attached to a 10-ft (3 m) microcable that permits a door to close on it. Mount meter with included hook-and-loop strips, wall-mount slots, or use flip-open stand on benchtop.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
29,000.00 - ₹70,100.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° F) No. of Probe(s) Included Number Of Probe Channels
Availability Pricing
Traceable Memory-Loc™ Datalogging Thermometer; 1 Glass Bottle Probe 9446037 Mfr # 6440TR
COMPARE
-58 1 1
₹30,700.00INR / Each
Traceable Memory-Loc™ Datalogging Thermometer; 2 Glass Bottle Probes 9446039 Mfr # 6441TR
COMPARE
-58 2 2
₹32,800.00INR / Each
Traceable Memory-Loc™ Datalogging Thermometer; 1 Bullet Probe 9446041 Mfr # 6442TR
COMPARE
-58 1 1
₹29,000.00INR / Each
Traceable Memory-Loc™ Datalogging Thermometer; 2 Bullet Probes 9446043 Mfr # 6443TR
COMPARE
-58 2 2
₹32,300.00INR / Each
Traceable Memory-Loc™ Datalogging Thermometer; 1 Stainless Steel Probe 9446044 Mfr # 6445TR
COMPARE
-58 1 1
₹33,700.00INR / Each
Traceable Memory-Loc™ Datalogging Thermometer; 2 Stainless Steel Probes 9446046 Mfr # 6446TR
COMPARE
-58 2 2
₹36,500.00INR / Each
Traceable Memory-Loc™ Datalogging Thermometer; 1 Glass Bottle/1 Bullet Probe 9446047 Mfr # 6444TR
COMPARE
-58 2 2
₹33,700.00INR / Each
Traceable Memory-Loc™ Datalogging Thermometer; 1 Vaccine Bottle Probe 9876765 Mfr # 6447TR
COMPARE
-58 1 1
₹30,500.00INR / Each
Traceable Memory-Loc™ Datalogging Thermometer; 2 Vaccine Bottle Probes 9876766 Mfr # 6448TR
COMPARE
-58 2 2
₹32,800.00INR / Each
Traceable Memory-Loc™ Datalogging Low-Temp Thermometer; 1 Stainless Steel Probe 9876855 Mfr # 6428TR
COMPARE
-130 1 1
₹40,700.00INR / Each
Traceable Memory-Loc™ Datalogging Low-Temp Thermometer; 2 Stainless Steel Probes 9876856 Mfr # 6429TR
COMPARE
-130 2 2
₹43,700.00INR / Each
Traceable Memory-Loc™ Datalogging Cryogenic Thermometer; 1 Stainless Steel Probe 9876859 Mfr # 6458TR
COMPARE
-328 1 1
₹63,600.00INR / Each
Traceable Memory-Loc™ Datalogging Cryogenic Thermometer; 2 Stainless Steel Probes 9876860 Mfr # 6459TR
COMPARE
-328 2 2
₹70,100.00INR / Each
Showing 1- 13 of 13