Traceable® Data Logging Thermometer Bundles with TraceableLIVE® Subscription
One year standard TraceableLIVE® subscription included with purchase
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • TraceableLIVE-capable products securely monitor your critical environments and notify your smartphone, tablet, or PC if anything goes wrong
  • Notifications for peace of mind - everyone on your team can receive e-mail, text, and push notifications on the environments you monitor
  • Remotely view data logging history, effortlessly generate reports, and control alarm parameters
  • Digital data reports sent with ease whenever you need them
  • Customize your report with location, device, date range, and report format
Order a thermometer for your unique application and get a one-year standard TraceableLIVE® subscription included.
A standard subscription includes the following features: E-mail and Push Alarms, Unlimited Data Storage, Unlimited Devices, Unlimited Team Members, and Calibration Due notification. Optional SMS text notifications are not included with the subscription code, but can be added on for (+2$/device/month) in the TraceableLIVE® app.
Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.
70,600.00 - ₹90,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Input Type Max Temperature (° C) Max Temperature (° F)
Availability Pricing
Traceable Wi-Fi Data Logging Refrigerator/Freezer Thermometer Bundle, 2 Bottle Probes​, with 1-Year Standard TraceableLIVE® Subscription 6500260 Mfr # 6655TR
COMPARE
Two bottle probes 60 140
₹70,600.00INR / Each
Traceable Wi-Fi Data Logging Ultra-Low Temperature Thermometer Bundle, 1 Stainless Steel Probe​, with 1-Year Standard TraceableLIVE® Subscription 6500261 Mfr # 6656TR
COMPARE
One stainless steel RTD probe 105 221
₹90,900.00INR / Each