Traceable 2-in-1 Waterproof Food HACCP Thermometer; 1 Integral Fold-Out Probe

Representative image only.

Traceable 2-in-1 Waterproof Food HACCP Thermometer; 1 Integral Fold-Out Probe

Traceable | Mfr: 4364TR| Item: 9876737

Designed specifically as a HACCP (Hazard Analysis Critical Control Point) thermometer for food safety

₹27,400.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

₹27,400.00INR / Each

Buy It Together

Current Item

Traceable 2-in-1 Waterproof Food HACCP Thermometer; 1 Integral Fold-Out Probe
Traceable 2-in-1 Waterproof Food HACCP Thermometer; 1 Integral Fold-Out Probe
9876737

₹27,400.00INR / Each
+ + + + + +
Cole-Parmer RTD Thermometer, NIST-Traceable Calibration
2025095

₹24,800.00INR / Each
+ =
Total Price 204800.00 ADD 8 ITEMS TO CART
Product Description

Includes

Traceable® Certificate, integral fold-out probe, hook-and-loop strip, and batteries.

Key Features

  • Noncontact infrared and 6"L piercing stainless steel probe combine for versatility
  • Waterproof case and unique membrane keys prevent splashes from permeating unit
  • Readings update every second
  • Adjustable emissivity from 0.1 to 1.0
  • Distance-to-target size ratio is 2.5:1
  • Includes NIST-traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • 1 year warranty

More About this Item

Ensure food safety with this food HACCP thermometer. LEDs indicate safe cool/frozen, safe holding, or danger zone conditions at a quick glance; green light under 4°C (40°F) or over 60°C (140°F) and red danger light between 4 to 60°C (40 to 140°F).

Easily measure temperatures of liquids and semi-solids using the piercing stainless steel probe. An infrared thermometer allows temperature measurements from a distance. Waterproof construction allows for use in labs, plants, and all food areas—simply wash down to clean. Meter features Min/Max readings, selectable °F/°C, and low-battery indicator.

Traceable® Products come with an individually numbered Traceable® Certificate provided with each unit, assuring accuracy from our ISO/IEC 17025:2017 (1750.01) calibration laboratory, accredited by A2LA. The certificate indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMIs) that are signatories to the CIPM Mutual Recognition Agreement. Every Traceable® product undergoes an intense vetting process before it can carry the Traceable name, saving you both time and money by eliminating the need for separate calibration.

Guaranteed to have at least one year of calibration remaining on certificate.

Cole-Parmer provides a wide offering of Professional Infrared Thermometers with varying features and accuracies depending on your application.

However, none of our infrared thermometers, no matter the manufacture, are approved for medical usage, and are not FDA approved.

For help in selecting an infrared thermometer for industrial, food or laboratory usage please contact the customer service team at Cole-Parmer.

1 Year WarrantyConformite EuropeenneNIST Traceable Calibration
Specifications
  • Meter Measurement TypeDigital Indicator and Infrared
  • Number Of Probe Channels1
  • Min Temperature (° F)-67
  • Max Temperature (° F)626
  • Min Temperature (° C)-55
  • Max Temperature (° C)330
  • Accuracy±0.5°C from -5 to 65°C, ±1.0°C below -5°C, ±1% above 65°C
  • Resolution0.2°C (0.5°F) between –10 and 200° otherwise 1°
  • Min IR Temperature (° F)-67
  • Min IR Temperature (° C)-55
  • Max IR Temperature (° F)482
  • Max IR Temperature (° C)250
  • IR Temperature Accuracy±2% or 2°C
  • IR Temperature Resolution (° C)0.2°C (0.5°F) between –10 and 200°; otherwise 1°
  • IP RatingIP65
  • Min/Max FunctionYes, available for IR thermometer only
  • Low Battery IndicatorYes
  • Display TypeLCD, 1/4"H digits
  • Height (in)1 1/2
  • Width (in)1
  • Length (in)6 1/2
  • No. of Probe(s) Included1
  • Probe Diameter (in)0.125
  • Probe Length (in)6
  • Battery2 AAA (included)
  • Probe TypePiercing tip
  • Description2-in-1 Waterproof Food HACCP Thermometer; 1 Integral Fold-Out Probe
  • Warranty1 year
Reviews
Q&A