Cole-Parmer RTD Thermometer, NIST-Traceable Calibration
Representative image only.
Cole-Parmer RTD Thermometer, NIST-Traceable Calibration
Cole-Parmer | Item: 2025095
Perfect for the lab to production floor
₹24,800.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹24,800.00INR / Each
Buy It Together
Current Item
Cole-Parmer RTD Thermometer, NIST-Traceable Calibration
Cole-Parmer RTD Thermometer, NIST-Traceable Calibration
2025095
₹24,800.00INR / Each
+ + + + + + + + =
Total Price 279600.00 ADD 9 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Protective rubber armor boot with built-in stand, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal. Data logging models 20250-92, -94, and -96 also include a USB cable and software.
Key Features
  • Reliable measurements, guaranteed!
  • NIST-traceable certificate included
  • Heavy duty construction to protect against knocks and jars.
  • Super large LCD for easy viewing of data
  • Accepts a wide variety of probes
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
  • 2 year warranty
More About this Item
The Cole-Parmer® Professional Single-Input Thermometers offer fast response and laboratory accuracy. Features include automatic backlight, Max/Min readings, and automatic power-off. These thermometers provide precise measurements when accuracy is a must, delivering dependable results every time! Each model is packed with distinctive features for use in a wide array of applications. Robust and reliable constructed to protects against knocks and jars. These thermometers produce a high degree of accuracy, with a rapid response time, and exceptional battery life of up to 3000 hours.
NIST Traceable CalibrationTwo year warranty
Specifications
  • Meter Measurement TypeRTD
  • Number Of Probe Channels1
  • Types Of Probes AcceptedRTD
  • Min Temperature (° F)-320
  • Max Temperature (° F)1562
  • Min Temperature (° C)-200
  • Max Temperature (° C)850
  • Accuracy±1.5⁰F (0.8⁰C) from -148 to 392⁰F (-99.9 to 199.9⁰C); ±2⁰F (1⁰C) ±0.5% of reading outside range
  • Resolution0.1⁰F (0.1⁰C) from 148 to 1562⁰F (-99.9 to 850⁰C); 1⁰F (1⁰C) outside range
  • Data LoggingNo
  • Low Battery IndicatorYes
  • Display TypeLarge, dual, backlit LCD
  • Battery Life300 hours
  • Battery1 9V (included)
  • DescriptionRTD Thermometer, NIST Traceable Calibration