Thermo Scientific 9409BN Solid State Half-Cell ISE Electrode; Fluoride
Representative image only.
Thermo Scientific 9409BN Solid State Half-Cell ISE Electrode; Fluoride
Thermo Scientific | Mfr: 9409BN| Item: 0572111
For a complete system:
₹168,000.00INR / Each
₹168,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific 9409BN Solid State Half-Cell ISE Electrode; Fluoride
Thermo Scientific 9409BN Solid State Half-Cell ISE Electrode; Fluoride
0572111
₹168,000.00INR / Each
+ + + =
Total Price 451300.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Ion-selective electrode
  • Reference electrode
  • Standardizing solution
  • Filling solution for reference electrodes
  • Ionic strength and pH adjustors
  • 3 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Select the ion-selective electrodes, reference electrodes, and necessary solutions to best fit your application. Choose ion-selective electrodes with BNC connectors and 3-ft cord. Note: All electrodes listed are sensing half-cells which require a reference electrode.
Specifications
  • ApplicationsHalf-cell
  • Electrode TypeSolid-state
  • Reference JunctionSure-Flow
  • Reference ElectrolyteOptimum Results A electrode fi lling solution
  • Ion-Selective ElectrodeFluoride
  • Min pH (pH)5
  • Max pH (pH)7
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • DescriptionSolid State Half-Cell ISE Electrode; Fluoride
  • Warranty3 Month Product / 12 Month Parts and Labor