Thermo Scientific 940911 Ionic strength and pH adjustors, TISAB III (conc.)
Representative image only.
Thermo Scientific 940911 Ionic strength and pH adjustors, TISAB III (conc.)
Thermo Scientific | Mfr: 940911| Item: 0573016
Use these ionic strength and pH adjustors for optimal performance of your ISE
₹41,000.00INR / Each
₹41,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific 940911 Ionic strength and pH adjustors, TISAB III (conc.)
Thermo Scientific 940911 Ionic strength and pH adjustors, TISAB III (conc.)
0573016
₹41,000.00INR / Each
+ =
Total Price 41000.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Calibrate your instrument and help perform addition and subtraction operations
  • 3 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Combine with ionic strength and pH adjusters to eliminate interferences from other ion species and adjust standards and samples to an optimal pH range. Standardizing and filling solutions as well as ion-selective electrodes are sold separately.
Specifications
  • Ion-Selective ElectrodeFluoride
  • DescriptionIonic strength and pH adjustors, TISAB III (conc.)
  • Warranty3 Month Product / 12 Month Parts and Labor