Thermo Scientific 940909 Ionic Strength and pH Adjustors, TISAB II, 10 ppm; 1 Gallon
Representative image only.
Thermo Scientific 940909 Ionic Strength and pH Adjustors, TISAB II, 10 ppm; 1 Gallon
Thermo Scientific | Mfr: 940909| Item: 0573015
Use these ionic strength and pH adjustors for optimal performance of your ISE
₹41,800.00INR / Gallon of 1
₹41,800.00INR / Gallon of 1
Buy It Together
Current Item
Thermo Scientific 940909 Ionic Strength and pH Adjustors, TISAB II, 10 ppm; 1 Gallon
Thermo Scientific 940909 Ionic Strength and pH Adjustors, TISAB II, 10 ppm; 1 Gallon
0573015
₹41,800.00INR / Gallon of 1
+ =
Total Price 63300.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Calibrate your instrument and help perform addition and subtraction operations
  • 3 Month Product / 12 Month Parts and Labor Warranty
More About this Item
Combine with ionic strength and pH adjusters to eliminate interferences from other ion species and adjust standards and samples to an optimal pH range. Standardizing and filling solutions as well as ion-selective electrodes are sold separately.
Specifications
  • Ion-Selective ElectrodeFluoride
  • DescriptionIonic strength and pH adjustors, TISAB II
  • Warranty3 Month Product / 12 Month Parts and Labor