Thermo Scientific 900100 Sure-Flow Reference Electrode, SJ; Pin
Representative image only.
Thermo Scientific 900100 Sure-Flow Reference Electrode, SJ; Pin
Thermo Scientific | Mfr: 900100| Item: 0571001
Get precision measurements of virtually any sample
₹52,000.00INR / Each
₹52,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific 900100 Sure-Flow Reference Electrode, SJ; Pin
Thermo Scientific 900100 Sure-Flow Reference Electrode, SJ; Pin
0571001
₹52,000.00INR / Each
+ =
Total Price 78800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Great for applications where rough handling and breakage are a problem Use in conjunction with a separate reference electrode
  • 3 month warranty
More About this Item
Electrodes combine the Sure-Flow easy-clean junction with the speed, accuracy, and stability of Ross electrodes.
When filled with Orion 05730-38 Filling Solution, the Orion 05710-01 matches the potential characteristics of a conventional KCl saturated calomel electrode. Reference electrode Orion 05710-01 is recommended for all pH and ion-selective measurements except for those:
In dilute solutions where the ion to be measured is contained in the filling solution of the Orion 05710-01
In solutions where an ion in the filling solution would introduce an electrode interference
In solutions where a species is present which forms a precipitate with an ion contained in the filling solution of the Orion 05710-01
For these incompatible samples, a double-junction electrode, like the 05710-10, is recommended.
3 Month Warranty
Specifications
  • ApplicationsHalf cell, reference
  • Electrode TypeSingle junction
  • Electrode Body MaterialEpoxy
  • Reference JunctionSure-flow
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • ConnectorPin-tip (U.S. standard)
  • Cable Length (m)1
  • DescriptionSure-Flow Reference Electrode, SJ; Pin