Supco SL500T Temperature Data Logger with Real Time LCD
Representative image only.
Supco SL500T Temperature Data Logger with Real Time LCD
Supco | Mfr: SL500T| Item: 1800052
USB interface for fast data transfer
Call For Price
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Supco SL500T Temperature Data Logger with Real Time LCD
Supco SL500T Temperature Data Logger with Real Time LCD
1800052
Call For Price
+ + + =
Total Price 56500.00 ADD 4 ITEMS TO CART
Product Description
Includes
Windows®-based software, 3-ft (0.9 m) USB interface cable, and battery. Models 18000-54 and 18000-56 also include an external probe.
Key Features
  • Real-time display of temperature, relative humidity (SL500TH only), time, date, alarm status, logging status, and battery life
  • Nonvolatile memory retains collected data—even if the battery fails
  • 1 year warranty
More About this Item
Supco SL500 data loggers are compact, light weight, and easy to use—making them perfect for many applications. Each logger includes software needed for logger set up, data downloading, and analysis. Loggers can be programmed to record over points once the memory is full, or stop recording. Sampling rates can be adjusted from every 1 second to 9 hours, and start/stop may be manually controlled on the logger or a specific run time may be set through the software. The temperature and relative humidity model (18000-53) alternates display of temperature and humidity every two seconds.
1 Year WarrantyConformite Europeenne
Specifications
  • Number of Channels1
  • Input TypeTemperature
  • Max Temperature (° F)160
  • Min Temperature (° F)-5
  • Max Temperature (° C)70
  • Min Temperature (° C)-20
  • Temperature Accuracy (° F)±0.9
  • Temperature Accuracy (° C)±0.5
  • Memory Size43344 datapoints
  • Battery Life3 years average use
  • Battery Type1 3V CR2032 lithium (included)
  • Display TypeLCD
  • DescriptionTemperature Data Logger with Real Time LCD