Traceable® Temperature and Temperature/Humidity Bluetooth® Data Loggers Compatible with TraceableGO™ App
Easily access collected data between destinations with Bluetooth® data loggers
  • Connect to your data via smartphone or tablet—any Bluetooth-enabled device—using a downloadable Bluetooth App
  • View and download data with free TraceableGO™ App - data can be exported from the TraceableGO™ app and loaded to TraceableLIVE® cloud service*
  • Data loggers store up to 90 alarm events
  • Readings and setup parameters easily viewable on large 2" by 1" LCD
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Track environmental conditions of reagents, clinical samples, vaccines, pharmaceuticals, and food items during transport to any location—Bluetooth® loggers monitor shipments while in transit. Compact, durable data loggers fit almost anywhere—even inside a small cooler.
Configure the data loggers from your device using the TraceableGO™ App to start; stop; change memory mode, unit preference, alarm setting, logging interval; and enable/disable alarm. When all the parameters are set, simply press START on the logger to begin monitoring. Once the shipment is delivered, double press START/STOP on the logger to enable Bluetooth® and export the data to your device. You will get an immediate response to see if your contents have been exposed to conditions outside the set parameters.
Measurements are updated every 5 seconds with user-defined reading intervals from 1 minute to 12 hours in 1-minute steps. Logging duration is 227 days (7.5 months) on a 5-minute logging interval. Data loggers store up to 90 alarm events. Data logger displays current reading, Min/Max, and run time or alarm time. 18004-13 only: Display toggles between temperature and humidity measurements every 5 seconds.
You have the option to subscribe to cloud-based TraceableLIVE® service to upload data to cloud—ideal for tracking data from multiple loggers or to comply with regulatory agency requirements. The low-cost TraceableLIVE® subscription provides unlimited trip data to be saved on the cloud as well as the ability to send a report to an email.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Related Article: How to Safely Transport Vaccines, how to meet the critical challenge of ensuring a temperature-controlled supply chain from manufacturer to end-user.
9,200.00 - ₹24,800.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Input Type Max Temperature (° C) Min Temperature (° C)
Availability Pricing
COMPARE
Temperature; integrated sensor 70 -20
₹9,200.00INR / Each
COMPARE
Temperature and humidity; integrated sensors 70 -20
₹9,400.00INR / Each
COMPARE
Temperature; bullet probe 70 -50
₹10,500.00INR / Each
COMPARE
Temperature; bottle probe 70 -50
₹20,000.00INR / Each
COMPARE
Temperature; platinum probe 100 -90
₹24,800.00INR / Each