Spectra Por 131348 Biotech-Grade CE Dialysis Tubing, 20000 MWCO, 31mm/20mm; 33ft

Representative image only.

Spectra Por 131348 Biotech-Grade CE Dialysis Tubing, 20000 MWCO, 31mm/20mm; 33ft

Spectra Por | Mfr: 131348| Item: 0289012

Ideal for concentration, purification, and desalination

₹134,000.00INR / Box of 1

₹134,000.00INR / Box of 1

Buy It Together

Current Item

Spectra Por 131348 Biotech-Grade CE Dialysis Tubing, 20000 MWCO, 31mm/20mm; 33ft
Spectra Por 131348 Biotech-Grade CE Dialysis Tubing, 20000 MWCO, 31mm/20mm; 33ft
0289012

₹134,000.00INR / Box of 1
+ + + =
Total Price 245000.00 ADD 4 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • High-purity membranes with no heavy metal or sulfide contaminants
  • Low protein adsorption
  • 1 year warranty

More About this Item

Cellulose Ester (CE) offers a wider range of more precise MWCO’s for critical separations; packaged as a 10-m roll in 0.05% sodium azide.

1 Year Warranty
Specifications
  • MaterialCellulose Ester, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)20000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)20000
  • Flat Width (mm)31
  • Diameter (mm)20
  • Length Per Pack (ft)33
  • Diameter (in)0.787
  • DescriptionBiotech-Grade CE Dialysis Tubing, 20000 MWCO, 31mm/20mm; 33ft
  • Warranty1 year
Reviews
Q&A