Spectra/Por Float-A-Lyzer G2 Dialysis Device
Disposable ready-to-use dialysis device eases sample loading and retrieval
  • 95 to 98% sample recovery, 99% sample purity and <10% sample dilution
  • Leak-proof screw cap allows easy pipette access for sample loading, testing, and retrieval
  • Ideal for dialyzing electro-eluted proteins and chromatography fractions
The Float-A-Lyzer G2 features biotech grade cellulose ester (CE) membrane. The low protein-binding, ultrapure membrane (no heavy metals and sulfides) is available in nine precise MWCO''s from 100 to 1,000,000 Daltons. The color-coded, re-sealable screw cap eliminates leaks associated with knots, closures, and needle punctures and allows repeated access for in-process testing.
The cylindrical tubing geometry prevents sample dilution and provides open access for total volume retrieval with included pipette. The included floatation ring and weighted bottom maintain vertical orientation during dialysis. This design also permits several units to fit in a single reservoir. All sizes are supplied dry with glycerol in a package of 12.
41,000.00 - ₹56,000.00INR / Pkg of 12
26 variations of this product are available.
See All Product Options Below
26 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Molecular Weight Cut Off (Mwco) (Dalton) Min Molecular Weight Cut Off (Mwco) (Dalton) Sample Volume (µL)
Availability Pricing
Spectra Por G235025 Float-A-Lyzer G2 Dialysis Device, 100-500 MWCO, 1 mL; 12/Pk 0290620 Mfr # G235025
COMPARE
500 100 1
₹41,000.00INR / Pkg of 12
Spectra Por G235027 Float-A-Lyzer G2 Dialysis Device, 500-1000 MWCO, 1 mL; 12/Pk 0290621 Mfr # G235027
COMPARE
1000 500 1
₹41,000.00INR / Pkg of 12
Spectra Por G235029 Float-A-Lyzer G2 Dialysis Device, 3500-5000 MWCO, 1 mL; 12/Pk 0290622 Mfr # G235029
COMPARE
5000 3500 1
₹41,000.00INR / Pkg of 12
Spectra Por G235031 Float-A-Lyzer G2 Dialysis Device, 8000-10000 MWCO, 1 mL; 12/Pk 0290623 Mfr # G235031
COMPARE
10000 8000 1
₹41,000.00INR / Pkg of 12
Spectra Por G235033 Float-A-Lyzer G2 Dialysis Device, 20000 MWCO, 1 mL; 12/Pk 0290624 Mfr # G235033
COMPARE
20000 20000 1
₹41,000.00INR / Pkg of 12
Spectra Por G235034 Float-A-Lyzer G2 Dialysis Device, 50000 MWCO, 1 mL; 12/Pk 0290625 Mfr # G235034
COMPARE
50000 50000 1
₹41,000.00INR / Pkg of 12
Spectra Por G235035 Float-A-Lyzer G2 Dialysis Device, 100000 MWCO, 1 mL; 12/Pk 0290626 Mfr # G235035
COMPARE
100000 100000 1
₹41,000.00INR / Pkg of 12
Spectra Por G235036 Float-A-Lyzer G2 Dialysis Device, 300000 MWCO, 1 mL; 12/Pk 0290627 Mfr # G235036
COMPARE
300000 300000 1
₹41,000.00INR / Pkg of 12
Spectra Por G235037 Float-A-Lyzer G2 Dialysis Device, 1,000,000 MWCO, 1 mL; 12/Pk 0290628 Mfr # G235037
COMPARE
1000000 1000000 1
₹41,000.00INR / Pkg of 12
Spectra Por G235051 Float-A-Lyzer G2 Dialysis Device, 500-1000 MWCO, 5 mL; 12/Pk 0290631 Mfr # G235051
COMPARE
1000 500 5
₹43,700.00INR / Pkg of 12
Spectra Por G235053 Float-A-Lyzer G2 Dialysis Device, 3500-5000 MWCO, 5 mL; 12/Pk 0290632 Mfr # G235053
COMPARE
5000 3500 5
₹43,700.00INR / Pkg of 12
Spectra Por G235055 Float-A-Lyzer G2 Dialysis Device, 8000-10000 MWCO, 5 mL; 12/Pk 0290633 Mfr # G235055
COMPARE
10000 8000 5
₹43,700.00INR / Pkg of 12
Spectra Por G235057 Float-A-Lyzer G2 Dialysis Device, 20000 MWCO, 5 mL; 12/Pk 0290634 Mfr # G235057
COMPARE
20000 20000 5
₹43,700.00INR / Pkg of 12
Spectra Por G235058 Float-A-Lyzer G2 Dialysis Device, 50000 MWCO, 5 mL; 12/Pk 0290635 Mfr # G235058
COMPARE
50000 50000 5
₹43,700.00INR / Pkg of 12
Spectra Por G235059 Float-A-Lyzer G2 Dialysis Device, 100000 MWCO, 5 mL; 12/Pk 0290636 Mfr # G235059
COMPARE
100000 100000 5
₹43,700.00INR / Pkg of 12
Spectra Por G235060 Float-A-Lyzer G2 Dialysis Device, 300000 MWCO, 5 mL; 12/Pk 0290637 Mfr # G235060
COMPARE
300000 300000 5
₹43,700.00INR / Pkg of 12
Spectra Por G235062 Float-A-Lyzer G2 Dialysis Device, 1,000,000 MWCO, 5 mL; 12/Pk 0290638 Mfr # G235062
COMPARE
1000000 1000000 5
₹43,700.00INR / Pkg of 12
Spectra Por G235061 Float-A-Lyzer G2 Dialysis Device, 100-500 MWCO, 10 mL; 12/Pk 0290640 Mfr # G235061
COMPARE
500 100 10
₹48,000.00INR / Pkg of 12
Spectra Por G235063 Float-A-Lyzer G2 Dialysis Device, 500-1000 MWCO, 10 mL; 12/Pk 0290641 Mfr # G235063
COMPARE
1000 500 10
₹48,000.00INR / Pkg of 12
Spectra Por G235065 Float-A-Lyzer G2 Dialysis Device, 3500-5000 MWCO, 10 mL; 12/Pk 0290642 Mfr # G235065
COMPARE
5000 3500 10
₹48,000.00INR / Pkg of 12
Showing 1 - 20 of 26