Spectra Por G235059 Float-A-Lyzer G2 Dialysis Device, 100000 MWCO, 5 mL; 12/Pk
Representative image only.
Spectra Por G235059 Float-A-Lyzer G2 Dialysis Device, 100000 MWCO, 5 mL; 12/Pk
Spectra Por | Mfr: G235059| Item: 0290636
Disposable ready-to-use dialysis device eases sample loading and retrieval
₹43,700.00INR / Pkg of 12
₹43,700.00INR / Pkg of 12
Buy It Together
Current Item
Spectra Por G235059 Float-A-Lyzer G2 Dialysis Device, 100000 MWCO, 5 mL; 12/Pk
Spectra Por G235059 Float-A-Lyzer G2 Dialysis Device, 100000 MWCO, 5 mL; 12/Pk
0290636
₹43,700.00INR / Pkg of 12
+ =
Total Price 144700.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 95 to 98% sample recovery, 99% sample purity and <10% sample dilution
  • Leak-proof screw cap allows easy pipette access for sample loading, testing, and retrieval
  • Ideal for dialyzing electro-eluted proteins and chromatography fractions
  • 1 year warranty
More About this Item
The Float-A-Lyzer G2 features biotech grade cellulose ester (CE) membrane. The low protein-binding, ultrapure membrane (no heavy metals and sulfides) is available in nine precise MWCO''s from 100 to 1,000,000 Daltons. The color-coded, re-sealable screw cap eliminates leaks associated with knots, closures, and needle punctures and allows repeated access for in-process testing.
The cylindrical tubing geometry prevents sample dilution and provides open access for total volume retrieval with included pipette. The included floatation ring and weighted bottom maintain vertical orientation during dialysis. This design also permits several units to fit in a single reservoir. All sizes are supplied dry with glycerol in a package of 12.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.
1 Year Warranty
Specifications
  • MaterialCellulose Ester, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)100000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)100000
  • Sample Volume (µL)5
  • Max Sample Volume (mL)5
  • ColorBlue
  • DescriptionFloat-A-Lyzer G2 Dialysis Device, 100000 MWCO, 5 mL; 12/Pk