The product you searched is discontinued. Below is the suggested replacement. Compare to discontinued item.

Spectra Por G235033 Float-A-Lyzer G2 Dialysis Device, 20000 MWCO, 1 mL; 12/Pk

Representative image only.

Spectra Por G235033 Float-A-Lyzer G2 Dialysis Device, 20000 MWCO, 1 mL; 12/Pk

Spectra Por | Mfr: G235033| Item: 0290624

Disposable ready-to-use dialysis device eases sample loading and retrieval

₹41,000.00INR / Pkg of 12

₹41,000.00INR / Pkg of 12

Buy It Together

Current Item

Spectra Por G235033 Float-A-Lyzer G2 Dialysis Device, 20000 MWCO, 1 mL; 12/Pk
Spectra Por G235033 Float-A-Lyzer G2 Dialysis Device, 20000 MWCO, 1 mL; 12/Pk
0290624

₹41,000.00INR / Pkg of 12
+ =
Total Price 142000.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • 95 to 98% sample recovery, 99% sample purity and <10% sample dilution
  • Leak-proof screw cap allows easy pipette access for sample loading, testing, and retrieval
  • Ideal for dialyzing electro-eluted proteins and chromatography fractions
  • 1 year warranty

More About this Item

The Float-A-Lyzer G2 features biotech grade cellulose ester (CE) membrane. The low protein-binding, ultrapure membrane (no heavy metals and sulfides) is available in nine precise MWCO''s from 100 to 1,000,000 Daltons. The color-coded, re-sealable screw cap eliminates leaks associated with knots, closures, and needle punctures and allows repeated access for in-process testing.
The cylindrical tubing geometry prevents sample dilution and provides open access for total volume retrieval with included pipette. The included floatation ring and weighted bottom maintain vertical orientation during dialysis. This design also permits several units to fit in a single reservoir. All sizes are supplied dry with glycerol in a package of 12.

Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.

1 Year Warranty
Specifications
  • MaterialCellulose Ester, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)20000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)20000
  • Sample Volume (µL)1
  • Max Sample Volume (mL)1
  • ColorRed
  • DescriptionFloat-A-Lyzer G2 Dialysis Device, 20000 MWCO, 1 mL; 12/Pk
  • Warranty1 year
Reviews
Q&A