SP Hotpack Spindle Rack Set for Glassware Washer, Full Upper and Lower Spindle Racks
Representative image only.
SP Hotpack Spindle Rack Set for Glassware Washer, Full Upper and Lower Spindle Racks
| Mfr: 00519289| Item: 7890363
Accessories for SP Hotpack® glassware washers
₹1,430,000.00INR / Each
₹1,430,000.00INR / Each
Buy It Together
Current Item
SP Hotpack Spindle Rack Set for Glassware Washer, Full Upper and Lower Spindle Racks
SP Hotpack Spindle Rack Set for Glassware Washer, Full Upper and Lower Spindle Racks
7890363
₹1,430,000.00INR / Each
+ + + + +
Labconco 4587000 Standard upper rack for Glassware Washers
9896360
₹197,000.00INR / Each
=
Total Price 11400000.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Optimize performance of glassware washers with accessories such as rack inserts
  • Suitable for use with for SP Hotpack® vertical, undercounter, and freestanding glassware washers
  • Full upper and full lower racks feature 44 spindles total
  • Use lower rack for taller glassware
  • For use with spindle rack ready washers only
  • 1 year warranty
More About this Item
Choose from a wide range of accessories to enhance performance of your SP Hotpack® glassware washers. Accessories are available for use with the vertical, undercounter, or freestanding glassware washers, choose accessories based on the washer you have.
Baskets, insert racks, and spindle racks maximize space, improve efficiency, and protect specialty glassware. All insert accessories feature stainless steel construction for long life and durability.
1 Year Warranty
Specifications
  • MaterialStainless Steel
  • Compatible WithSP Hotpack spindle rack ready glassware washers
  • DescriptionSpindle Rack Set for Glassware Washer, Full Upper and Lower Spindle Racks