Labconco 4587000 Standard upper rack for Glassware Washers
Representative image only.
Labconco 4587000 Standard upper rack for Glassware Washers
Labconco | Mfr: 4587000| Item: 9896360
Optimize performance of your Labconco glassware washer
₹197,000.00INR / Each
₹197,000.00INR / Each
Buy It Together
Current Item
Labconco 4587000 Standard upper rack for Glassware Washers
Labconco 4587000 Standard upper rack for Glassware Washers
9896360
₹197,000.00INR / Each
+ +
SP Hotpack DI Water Boost Pump for Glassware Washer
7890362
₹228,000.00INR / Each
+ + + =
Total Price 10198000.00 ADD 6 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Adds more cleaning options to your washer
  • 1 year warranty
More About this Item
Choose from a wide range of rack accessories and other products to ensure top performance of your Labconco glassware washer.
Upper standard rack is included with each of the SteamScrubber® washers. Racks may be used in the FlaskScrubber® and FlaskScrubber® Vantage® washers as well. Constructed of 304 stainless steel, accommodates various inserts and racks to suit your glassware mix. Comes with release latches and wash arm.
1 Year Warranty
Specifications
  • DescriptionStandard upper rack for Glassware Washers