SP Hotpack® Undercounter and Freestanding Glassware Washers
Efficient, minimal water consumption with spotless results
  • Designed and insulated for quiet operation
  • Optimize temperature and time for specific cleaning with 6-cycle selection
  • Enhanced dual fan drying system draws in dry air and exhausts moist air out
  • Ensure long life and corrosion protection with 304 stainless steel chamber
  • Delayed start feature allows washer to operate unattended during off-peak hours to reduce energy costs
Designed for ease of operation and flexible loading, these SP Hotpack® undercounter and freestanding glassware washers feature upper and lower open racks and can accommodate a wide range of glassware. All models feature a soft touch controller, improved dryer, and noise buffering insulation for a quieter run. Three separate filters protect pumps from debris and are conveniently located for easy access. Multiple cycle selections and microprocessor control systems ensure that each load is processed with minimum energy consumption. Six cycle selections let you select optimal temperature and include a customizable timed option. These 6 cycles include rinse only, normal wash, daily wash, heavy wash, heavy wash time saver, and timed programs.
Choose from undercounter and freestanding models with standard, DI rinse, and DI rinse/spindle rack ready features. Undercounter units have adjustable height leveling feet and freestanding units have locking casters for portability. All DI models use facility-provided deionized water for final rinse and have the option of a DI booster pump (sold separately), recommended when the incoming DI water source has a pressure of less than 18 psi. Spindle-ready models can accommodate direct injection spindle racks (sold separately) for narrow-neck glassware.
1,535,000.00 - ₹3,026,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Exterior Depth (in) Exterior Height (in) Description
Availability Pricing
SP Hotpack Glassware Washer, Undercounter, with Tap Water Rinse, Top and Bottom Standard Racks; 115V 60 Hz 7890350 Mfr # UCW-HA00001
COMPARE
22.88 36.38 Glassware Washer, Undercounter, with Tap Water Rinse and Top and Bottom Standard Racks; 115V 60 Hz
₹1,535,000.00INR / Each
SP Hotpack Glassware Washer, Undercounter, with DI Rinse, Top and Bottom Standard Racks; 115V 60 Hz 7890351 Mfr # UCW-SA10001
COMPARE
22.88 36.38 Glassware Washer, Undercounter, with DI Rinse and Top and Bottom Standard Racks; 115V 60 Hz
₹2,078,000.00INR / Each
SP Hotpack Glassware Washer, Undercounter, with  DI Rinse, Top and Bottom Spindle Rack Ready; 115V 60 Hz 7890352 Mfr # UCW-SA20001
COMPARE
22.88 36.38 Glassware Washer, Undercounter, with DI Rinse and Top and Bottom Spindle Rack Ready; 115V 60 Hz
₹2,638,000.00INR / Each
SP Hotpack Glassware Washer, Freestanding, with Tap Water Rinse, Top and Bottom Standard Racks; 115V 60 Hz 7890354 Mfr # UCW-HA00101
COMPARE
24.25 38.5 Glassware Washer, Freestanding, with Tap Water Rinse and Top and Bottom Standard Racks; 115V 60 Hz
₹2,077,000.00INR / Each
SP Hotpack Glassware Washer, Freestanding, with DI Rinse, Top and Bottom Standard Racks; 115V 60 Hz 7890355 Mfr # UCW-HA10101
COMPARE
24.25 38.5 Glassware Washer, Freestanding, with DI Rinse and Top and Bottom Standard Racks; 115V 60 Hz
₹2,527,000.00INR / Each
SP Hotpack Glassware Washer, Freestanding, with  DI Rinse, Top and Bottom Spindle Rack Ready; 115V 60 Hz 7890356 Mfr # UCW-SA20101
COMPARE
24.25 38.5 Glassware Washer, Freestanding, with DI Rinse and Top and Bottom Spindle Rack Ready; 115V 60 Hz
₹3,026,000.00INR / Each