Sartorius Arium Mini Water Purification System with UV lamp

Representative image only.

Sartorius Arium Mini Water Purification System with UV lamp

Sartorius | Mfr: H2O-MM-UV-T-US| Item: 9929312

Eliminate tank cleaning with unique closed bag system

Call For Price

Call For Price

Buy It Together

Current Item

Sartorius Arium Mini Water Purification System with UV lamp
Sartorius Arium Mini Water Purification System with UV lamp
9929312

Call For Price
+
Sartorius H2O-S-PACK Arium® Scientific Pack Cartridge
9929331

₹127,000.00INR / Each
+ =
Total Price 127000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for small labs requiring <10 L/day of ultrapure water
  • Multicolor touch display with clear icons simplifies operation
  • Compact design with only a 11" (28 cm) wide footprint
  • 2 year warranty

More About this Item

The compact ultrapure water system features a closed bag tank system which protects pure water from secondary contamination and ensures consistent long-term high water quality. The 5-L bag can be quickly and easily replaced every six months, reducing maintenance time and eliminating the use of hazardous cleaning chemicals. System features include a favorites function to dispense recurring volumes, integrated pump for increasing water pressure, and pretreatment cartridge for effective purification.

Two models are available. The arium mini version is manually fed with pretreated water, while the arium mini plus version has direct connection to the portable water supply. Both are available with UV lamp (185/254 nm) to make sure the water is virtually free of organic components.

Consumables are needed to operate the unit and are sold separately. Please purchase a starter set along with your water system.

Two year warranty
Specifications
  • ASTM Water GradeType I
  • Max Flow Rate (L/min)1
  • Inlet Port0.25" OD tubing
  • With UV LampYes
  • With UltrafilterYes
  • SourceManual feeed with pre-treated water
  • Type I Water (Resistivity)18.2 MΩ-cm
  • Type I Water (TOC)≤5 ppb
  • Feed Water Requirements (TOC)<50 ppb
  • Type I Water (Conductivity)0.055 µS/cm
  • Type I Water (Bacteria)<1 CFU/1,000 mL
  • Feed Water Requirements (Temperature)2 to 30°C
  • Max Inlet Water Temperature (° C)30
  • Min Inlet Water Temperature (° C)2
  • Max Inlet Pressure (PSI)6
  • Min Inlet Pressure (bar)0.5
  • Power (VAC)100 to 240
  • Height (in)20.06
  • Depth (in)20.89
  • Width (in)11.02
  • Height (cm)50.94
  • Depth (cm)53.07
  • Width (cm)28
  • DescriptionWater Purification System with UV lamp
  • Warranty2 year
Reviews
Q&A