Sartorius Arium® Mini Essential Starter Set with UV Lamp

Representative image only.

Sartorius Arium® Mini Essential Starter Set with UV Lamp

Sartorius | Mfr: H2O-MU-UV-SET-US| Item: 9929449

Starter set includes all the required components to get you up and running

Call For Price

Call For Price

Buy It Together

Current Item

Sartorius Arium® Mini Essential Starter Set with UV Lamp
Sartorius Arium® Mini Essential Starter Set with UV Lamp
9929449

Call For Price
+ +
Sartorius H2O-CEL1 Arium® Mini UV Lamp, 185/254 nm
9929332

₹133,000.00INR / Each
=
Total Price 133000.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

Arium Mini Essential system with UV lamp, Scientific kit, 5 L bag, and SterilePlus filter.

Key Features

  • Ideal for small labs requiring <10 L/day of ultrapure water
  • Large touch display with self-explanatory navigation
  • Take up a small footprint—only measures 28 cm in width
  • Volume-controlled dispensing in 50 mL steps
  • Delivers up to 1 L/minute
  • Produces ASTM Type I ultrapure water
  • UV lamp effectively destroys organic compounds
  • 1 year warranty

More About this Item

Using high-quality water ensures the reproducibility of your results and prevents time-consuming repeat analysis. These compact Arium Mini systems only measure 28 cm (11") wide to easily fit on a benchtop. A high-resolution, touch-activated color display featuring self-explanatory icons is easy to navigate even while wearing gloves. Store favorite functions such repeated dispensing of identical volumes to help increase efficiency while minimizing errors.

Select from Arium Mini, Mini Plus, and Mini Essential depending on your application. All models produce up to 10 L of ASTM Type I ultrapure water per day. Arium Mini and Mini Plus use a closed Bagtank system while Mini Essential has no storage system. For analytical applications, order the Arium Mini with UV lamp to reduce total organic compound (TOC) to = 5 ppb.

Connect Arium Mini Essential water purification system to pretreated (RO, DI, or EDI) water line to produce ASTM Type I ultrapure water. Type I ultrapure water is useful in buffer, media, and reagent preparation and histology, ELISA, AAS, IC and photometry applications. The integrated UV lamp (185/254 nm) helps reduce total organic compound (TOC) to =5 ppb, making it ideal for analytical applications such as SPE, ICP-MS, GS-MS, LC-MS, and HPLC.

Conformite EuropeenneCanadian Standards Association
Specifications
  • ASTM Water GradeType I
  • Water Purification MethodAdsorption by spherical activated carbon, catalyst, reverse osmosis, ion exchange, optional UV irradiation, and by point-of-use particle-removing filtration
  • Max Flow Rate (L/min)1
  • Inlet Port0.25" OD tubing
  • With UV LampYes
  • SourcePretreated RO, DI, or EDI water line
  • Type I Water (Resistivity)18.2 MΩ × cm
  • Feed Water Requirements (Conductivity)< 100 μS/cm
  • Type I Water (TOC)≤ 5 ppb
  • Feed Water Requirements (TOC)< 50 ppb
  • Feed Water Requirements (turbidity)< 1 NTU
  • Type I Water (Conductivity)0.055 μS/cm
  • Type I Water (Bacteria)< 0.01 CFU/mL
  • Feed Water Requirements (pH range)4 to 10
  • Type I Water (Endotoxins)< 0.001 EU/mL
  • Feed Water Requirements (pressure)0 to 6.9 bar; (0 to approx. 100 psi); recommended > 2 bar (> 29 psi)
  • Feed Water Requirements (Temperature)2 to 30°C
  • Power (VAC)100 to 240
  • Height (in)20
  • Depth (in)20.9
  • Width (in)11
  • Height (cm)50.9
  • Depth (cm)53.1
  • Width (cm)28
  • DescriptionArium® Mini Essential Starter Set with UV Lamp
  • Warranty2 years
Reviews
Q&A