QLA Clip Style Basket for Dabigatran Etexilate Mesylate, 40 Mesh, 24.5 mm ID, 316 SS; Serialized
Representative image only.
QLA Clip Style Basket for Dabigatran Etexilate Mesylate, 40 Mesh, 24.5 mm ID, 316 SS; Serialized
| Mfr: CUSTBSK-40| Item: 4130859
QLA Baskets for specialized dissolution testing
₹91,600.00INR / Each
₹91,600.00INR / Each
Buy It Together
Current Item
QLA Clip Style Basket for Dabigatran Etexilate Mesylate, 40 Mesh, 24.5 mm ID, 316 SS; Serialized
QLA Clip Style Basket for Dabigatran Etexilate Mesylate, 40 Mesh, 24.5 mm ID, 316 SS; Serialized
4130859
₹91,600.00INR / Each
+ =
Total Price 190300.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of 316 electropolished stainless steel All baskets and shafts are laser marked with unique serial numbers
  • Use for Dabigatran Etexilate Mesylate
  • 24.5 mm ID basket
More About this Item
All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
Dabigatran Etexilate Mesylate requires a wider basket for dissolution testing. This oversize clip style basket is larger than the standard USP Apparatus 1 basket and will not fit on to a standard basket hub.
.
Specifications
  • Material316 stainless steel
  • Mesh Size40
  • DescriptionClip Style Basket for Dabigatran Etexilate Mesylate, 40 Mesh, 24.5 mm ID, 316 SS; Serialized