QLA Bent 316 SS Cannula for Hanson, 7.75"L; SS Luer Lock and Perm SS Tip
Representative image only.
QLA Bent 316 SS Cannula for Hanson, 7.75"L; SS Luer Lock and Perm SS Tip
| Mfr: CAN775-HR| Item: 4130248
Achieve consistent sampling with QLA Hanson-compatible probes and cannulas
₹54,600.00INR / Each
₹54,600.00INR / Each
Buy It Together
Current Item
QLA Bent 316 SS Cannula for Hanson, 7.75"L; SS Luer Lock and Perm SS Tip
QLA Bent 316 SS Cannula for Hanson, 7.75"L; SS Luer Lock and Perm SS Tip
4130248
₹54,600.00INR / Each
+
QLA Adjustable Cannula Stopper for Hanson; 1/8" (3.2 mm) OD
4130280
₹18,500.00INR / Each
=
Total Price 73100.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Matching cannula to stopper ensures greater sampling position consistency
  • 7.75" long with permanent SS tip
More About this Item
For dissolution tests, taking reliable samples is critical. QLA probes, cannula, and stoppers ensure consistent sampling position within the mixing vessel, which means more accurate testing.
Parts are made of 316 stainless steel that will not absorb or interfere with active ingredients.
.
Specifications
  • Material316 stainless steel
  • Length (")7 3/4
  • DescriptionBent 316 SS Cannula for Hanson, 7.75"L; SS Luer Lock and Perm SS Tip