QLA Adjustable Cannula Stopper for Hanson; 1/8" (3.2 mm) OD
Representative image only.
QLA Adjustable Cannula Stopper for Hanson; 1/8" (3.2 mm) OD
| Mfr: CANSTP-HR| Item: 4130280
QLA accessories help provide more precise and consistent dissolution results
₹18,500.00INR / Each
₹18,500.00INR / Each
Product Description
Key Features
  • For Hanson
  • 1/8" (3.2 mm) OD
More About this Item
Use stopper with cannula to ensure the sample is always taken from the same depth.
.
Specifications
  • Compatible WithHanson apparatus
  • DescriptionAdjustable Cannula Stopper for Hanson; 1/8" (3.2 mm) OD