Labnet D1302-230V AccuBlock™ Dry Block Heater, Dual; 230 VAC

Representative image only.

Labnet D1302-230V AccuBlock™ Dry Block Heater, Dual; 230 VAC

Labnet | Mfr: D1302-230V | Item: 5140122

Precise temperature regulation up to 150°C ensures accurate results

₹148,000.00INR / Each

₹148,000.00INR / Each

Buy It Together

Current Item

Labnet D1302-230V  AccuBlock™ Dry Block Heater, Dual; 230 VAC
Labnet D1302-230V AccuBlock™ Dry Block Heater, Dual; 230 VAC
5140122

₹148,000.00INR / Each
+ +
Labnet D1101 Block, solid for machining, no holes
3788499

₹29,000.00INR / Each
=
Total Price 199900.00 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Single, dual, and four block configurations available
  • Dual digital displays
  • External probe available for better traceability
  • 3 year warranty

More About this Item

AccuBlock Digital Dry baths are available in single, dual, and four block configurations. Ideal in a variety of applications including determining melting and boiling points, cell cultures, enzyme reactions and many more. Built of anodized, high grade non-porous aluminum, these units extremely durable and come with USB connectivity included.

3 Year WarrantyConformite EuropeenneCanadian Standards Association
Specifications
  • Number Of Blocks2
  • Length (")12 7/16
  • Width (InchShort)8 1/4
  • Height (InchShort)4 3/4
  • Uniformity (° C)0.2
  • Max Temperature (° F)302
  • Max Temperature (° C)150
  • Power (VAC)230
  • DescriptionLabnet D1302-230V AccuBlock™ Dry Block Heater, Dual; 230 VAC
  • Warranty3 year
Reviews
Q&A