AccuBlock Digital Dry Bath
Precise temperature regulation up to 150°C ensures accurate results
  • Single, dual, and four block configurations available
  • Dual digital displays
  • External probe available for better traceability
AccuBlock Digital Dry baths are available in single, dual, and four block configurations. Ideal in a variety of applications including determining melting and boiling points, cell cultures, enzyme reactions and many more. Built of anodized, high grade non-porous aluminum, these units extremely durable and come with USB connectivity included.
118,000.00 - ₹215,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Blocks Power (VAC) Description
Availability Pricing
Labnet D1301  AccuBlock™ Dry Block Heater, Single; 120 VAC 5140118 Mfr # D1301
COMPARE
1 120 Labnet D1301 AccuBlock™ Dry Block Heater, Single; 120 VAC
₹121,000.00INR / Each
Labnet D1302 AccuBlock™ Dry Block Heater, Dual; 120 VAC 5140119 Mfr # D1302
COMPARE
2 120 Labnet D1302 AccuBlock™ Dry Block Heater, Dual; 120 VAC
₹148,000.00INR / Each
Labnet D1304  AccuBlock™ Dry Block Heater, Quad; 120 VAC 5140120 Mfr # D1304
COMPARE
4 120 Labnet D1304 AccuBlock™ Dry Block Heater, Quad; 120 VAC
₹215,000.00INR / Each
Labnet D1301-230V  AccuBlock™ Dry Block Heater, Single; 230 VAC 5140121 Mfr # D1301-230V
COMPARE
1 230 Labnet D1301-230V AccuBlock™ Dry Block Heater, Single; 230 VAC
₹118,000.00INR / Each
Labnet D1302-230V  AccuBlock™ Dry Block Heater, Dual; 230 VAC 5140122 Mfr # D1302-230V
COMPARE
2 230 Labnet D1302-230V AccuBlock™ Dry Block Heater, Dual; 230 VAC
₹148,000.00INR / Each
Labnet D1304-230V  AccuBlock™ Dry Block Heater, Quad; 230 VAC 5140123 Mfr # D1304-230V
COMPARE
4 230 Labnet D1304-230V AccuBlock™ Dry Block Heater, Quad; 230 VAC
₹215,000.00INR / Each