Labnet D1196-PCR Single Block 96 Well PCR Plate
Representative image only.
Labnet D1196-PCR Single Block 96 Well PCR Plate
Labnet | Mfr: D1196-PCR| Item: 3788514
Uniformly molded high-grade aluminum blocks provide excellent temperature accuracy
₹70,100.00INR / Each
₹70,100.00INR / Each
Buy It Together
Current Item
Labnet D1196-PCR Single Block 96 Well Pcr Plate
Labnet D1196-PCR Single Block 96 Well Pcr Plate
3788514
₹70,100.00INR / Each
+ +
Labnet D1301  AccuBlock™ Dry Block Heater, Single; 120 VAC
5140118
₹121,000.00INR / Each
=
Total Price 405100.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Versatile range of interchangeable heating blocks work with many size sample tubes and plates
  • Heaters sold separately
  • PopStopper and AccuRack accessory sold seperately
  • 1 year warranty
More About this Item
Interchangeable aluminum heating blocks provide excellent thermal conductivity and accommodate various test tubes, cups, and plates. Thermometer wells available in all blocks except those used with plates.
1 Year Warranty
Specifications
  • Length (")3 1/100
  • Width (InchShort)3 35/64
  • Uniformity (° F)0.36
  • Uniformity (° C)0.2
  • Block Number Of Openings96
  • DescriptionLabnet AccuBlock D1196-PCR Single Block 96 Well PCR Plate