Labconco Purifier HEPA Filtered Enclosure, 4' W, 230 VAC

Representative image only.
Labconco Mfr # 3980422 – Item # WW-33507-22
Protect you from biological hazards requiring Biosafety Level 1, 2, and 3 containment
  • EPA filter removes 99.99% of particulates down to 0.3 µm
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Hood Width (in)48
  • Blower TypeStandard
  • LightsFluorescent
  • Exterior Width (in)47.75
  • Exterior Height (in)37.0625
  • Exterior Depth (in)28.6562
  • Power (VAC)230
  • Power (Hz)50
  • Exhaust Volume (CFM)220 to 310
  • DescriptionHEPA Filtered Enclosure, 4' W, 230V
  • Warranty1 Year
1 Year WarrantyETL ListedISO9001:2000 Certified Supplier

Key Features

  • EPA filter removes 99.99% of particulates down to 0.3 µm
  • 1 year warranty

More About this Item

Purifier enclosures protect you and your laboratory environment — they offer an economical alternative to Class II laminar flow cabinets when your applications do not require product protection. The filter indicator monitors the differential pressure across the filter and lets you know when to change the filter. Side panels and pivoting, angled sash are made of tempered safety glass; sash pivots upward 20" for easy cabinet loading. All units are shipped fully assembled. Order a spill-containing work surface and other accessories separately.

HEPA-Filtered Enclosures feature fluorescent lamp but do not include a UV light. They protect the user from hazardous chemical powders, dust, and allergens in diverse applications ranging from screening suspicious mail to weighing drug ingredients.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat