This product is now discontinued, View Related Products
Labconco Purifier Class I Biosafety Enclosures
Protect you from biological hazards requiring Biosafety Level 1, 2, and 3 containment
  • EPA filter removes 99.99% of particulates down to 0.3 µm
Purifier enclosures protect you and your laboratory environment—they offer an economical alternative to Class II laminar flow cabinets when your applications do not require product protection. The filter indicator monitors the differential pressure across the filter and lets you know when to change the filter. Side panels and pivoting, angled sash are made of tempered safety glass; sash pivots upward 20" for easy cabinet loading. All units are shipped fully assembled. Order a spill-containing work surface and other accessories separately.
Class I Enclosures protect the user from agents that require Biosafety Level 1, 2, or 3 containment but no product protection. These enclosures include an ultraviolet light that may be used in conjunction with surface disinfection to ensure thorough decontamination.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Exhaust Volume (CFM) Exterior Width (in) Power (VAC)
Availability Pricing
Labconco Purifier 3980203 Class 1 Filtered Enclosure, 2' W WW-33507-00 Mfr # 3980203
COMPARE
110 to 155 23.75 115
Call For Price
Labconco Purifier 3980223 Class 1 Filtered Enclosure, 2' W, 230 VAC WW-33507-02 Mfr # 3980223
COMPARE
110 to 155 23.75 230
Call For Price
Labconco Purifier 3980303 Class 1 Filtered Enclosure, 3' W, 115 VAC WW-33507-04 Mfr # 3980303
COMPARE
165 to 230 35.75 115
Call For Price
Labconco Purifier 3980323 Class 1 Filtered Enclosure, 3' W, 230 VAC WW-33507-06 Mfr # 3980323
COMPARE
165 to 230 35.75 230
Call For Price
Labconco Purifier Class 1 Filtered Enclosure, 4' W, 115 VAC WW-33507-08 Mfr # 3980403
COMPARE
220 to 310 47.75 115
Call For Price
Labconco Purifier 3980423 Class 1 Filtered Enclosure, 4' W, 230 VAC WW-33507-10 Mfr # 3980423
COMPARE
220 to 310 47.75 230