Labconco Fume Adsorbers

Trap harmful xylene and toluene fumes
Fume adsorbers are designed for light-duty applications involving organic solvents, formaldehyde, ammonia, or low molecular weight amines. Fume adsorbers use carbon filtration to trap vapors so that safe, filtered air is returned to the laboratory. No outside ducting is required. These portable, low-profile enclosures fit easily on countertops with overhanging cabinets and are available in two widths.
Each fume adsorber includes carbon filters that adsorbs vapors from organic solvents, formaldehyde or ammonia. Models 09003-40, -45, -75, -80, -85 and -90 require two filters; all other models require one filter. Features include built-in motorized impeller to provide face velocity up to 35 fpm. Adsorber has a white, dry powder epoxy-coated steel housing with perforated baffle for uniform airflow. Canopy is a clear, one-piece 3/8"-thick, shatter-resistant acrylic. Quiet operation is 58 to 63 dBA. A pre-wired rear-mounted auxiliary electrical receptacle has a top-mounted blower switch with circuit breaker. Work surface and base cabinet must be ordered separately.
What's included: Carbon filter (two for double models), power switch, and an 8-ft cord. The 115 VAC models include a three-prong plug.
671,000.00INR / Each

6 variations of this product are available.

See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Exterior Width (in) Power (VAC) Description
Availability Pricing
Labconco 3955220 Single-Filter Organic Solvent Fume Adsorber, 230V, 50/60 Hz 0900335 Mfr # 3955220
COMPARE
30 230 Single-Filter Organic Solvent Fume Adsorber, 230V, 50/60 Hz
₹671,000.00INR / Each
Labconco 3955420 Double-Filter Organic Solvent Fume Adsorber, 230V 0900345 Mfr # 3955420
COMPARE
60 230 Double-Filter Organic Solvent Fume Adsorber, 230V
Call For Price
Labconco 3955221 Single-filter Formaldehyde Fume Adsorber, 230V 0900365 Mfr # 3955221
COMPARE
30 230 Single-filter Formaldehyde Fume Adsorber, 230V
₹671,000.00INR / Each
Labconco 3955222 Single-filter Ammonia Fume Adsorber, 230V 0900370 Mfr # 3955222
COMPARE
30 230 Single-filter Ammonia Fume Adsorber, 230V
₹671,000.00INR / Each
Labconco 3955421 Double-filter Formaldehyde Fume Adsorber, 230V 0900385 Mfr # 3955421
COMPARE
60 230 Double-filter Formaldehyde Fume Adsorber, 230V
Call For Price
Labconco 3955422 Double-filter Ammonia Fume Adsorber, 230V 0900390 Mfr # 3955422
COMPARE
60 230 Double-filter Ammonia Fume Adsorber, 230V
Call For Price
Showing 1- 6 of 6

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat